Gene Transcripts
- Gene Symbol
- CSNK1G1
- Entrez Gene ID
- 53944
- Gene Name
- casein kinase 1 gamma 1
- Gene Aliases
- CK1gamma1
- Chromosome Location
- Chr. 15: 64165517 - 64356243 on Build GRCh38
- UniGene ID
- Hs.646508
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022048.3 |
NP_071331.2
|
8, 9 |
1340 |
|
GenBank mRNA |
AB042562.1 |
- |
7, 8 |
988 |
|
AB042563.1 |
- |
7, 8 |
972 |
|
AF223357.2 |
- |
7, 8 |
1058 |
|
AK026957.1 |
- |
4, 5 |
447 |
|
AK225960.1 |
- |
8, 9 |
1270 |
|
AL832551.1 |
- |
8, 9 |
1118 |
|
BC017236.2 |
- |
7, 8 |
789 |
|
BC130613.1 |
- |
8, 9 |
1113 |
40146644 |
BC136504.1 |
- |
8, 9 |
1113 |
9020493 |
View Full Product Details