Gene Transcripts
- Gene Symbol
- WDFY4
- Entrez Gene ID
- 57705
- Gene Name
- WDFY family member 4
- Gene Aliases
- C10orf64
- Chromosome Location
- Chr. 10: 48684815 - 48982956 on Build GRCh38
- UniGene ID
- Hs.287379
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020945.1 |
NP_065996.1
|
37, 38 |
6413 |
|
XM_005270004.3 |
XP_005270061.1
|
37, 38 |
6528 |
|
XM_011539986.2 |
XP_011538288.2
|
37, 38 |
6531 |
|
XM_011539987.2 |
XP_011538289.2
|
37, 38 |
6534 |
|
XM_011539988.2 |
XP_011538290.1
|
38, 39 |
6612 |
|
XM_011539990.2 |
XP_011538292.1
|
37, 38 |
6559 |
|
XM_017016463.1 |
XP_016871952.1
|
38, 39 |
6455 |
|
XM_017016464.1 |
XP_016871953.1
|
19, 20 |
2842 |
|
GenBank mRNA |
AB046827.1 |
- |
4, 5 |
596 |
|
AK074085.1 |
- |
17, 18 |
2471 |
|
View Full Product Details