Gene Transcripts

Gene Symbol
CBY3
Entrez Gene ID
646019
Gene Name
chibby family member 3
Gene Aliases
Chromosome Location
Chr. 5: 179677969 - 179681830 on Build GRCh38
UniGene ID
Hs.535767
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164444.1 NP_001157916.1 2 443
XM_017009726.1 XP_016865215.1 2 1296

View Full Product Details