Gene Transcripts

Gene Symbol
TGFBI
Entrez Gene ID
7045
Gene Name
transforming growth factor beta induced
Gene Aliases
BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1
Chromosome Location
Chr. 5: 136028895 - 136063818 on Build GRCh38
UniGene ID
Hs.369397
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000358.2 NP_000349.1 3 411
GenBank mRNA AK093916.1 - 3 266
AK094055.1 - 4 281
AK094581.1 - 3 355
AK222833.1 - 3 319
AK303643.1 - 3 315
BC000097.2 - 3 304 3506051
BC004972.1 - 3 299 2958878
BT009820.1 - 3 250
DA051240.1 - 3 411
KU178357.1 - 3 250
KU178358.1 - 3 250
M77349.1 - 3 297

View Full Product Details