Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
KCNE3 | NM_005472.4 | 3 | 2554 | NP_005463.1 |
XM_017017047.1 | 4 | 2624 | XP_016872536.1 | |
XM_017017048.1 | 5 | 2720 | XP_016872537.1 | |
XM_017017049.1 | 3 | 2570 | XP_016872538.1 | |
XM_017017050.1 | 3 | 5428 | XP_016872539.1 | |
XM_017017051.1 | 4 | 2543 | XP_016872540.1 | |
XM_017017052.1 | 3 | 2494 | XP_016872541.1 | |
BC113743.1 | 2 | 2378 | AAI13744.1 | |
CK301015.1 | 1 | 533 | ||
CR936625.1 | 4 | 2344 | CAI56768.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1041154 | Chr11:74403756 - 74466864 on Build GRCh38 | Loss | KCNE3 |
esv34979 | Chr11:74167643 - 74822407 on Build GRCh38 | Gain | POLD3 KCNE3 LOC101928580 PPME1 C2CD3 MIR4696 P4HA3 CHRDL2 RNF169 MIR548AL LOC100287896 LIPT2 PGM2L1 |
esv3626840 | Chr11:74371082 - 74464389 on Build GRCh38 | Gain | MIR548AL KCNE3 PGM2L1 |
esv2759837 | Chr11:74131564 - 74846552 on Build GRCh38 | Gain | POLD3 XRRA1 KCNE3 LOC101928580 PPME1 C2CD3 MIR4696 P4HA3 CHRDL2 RNF169 MIR548AL LOC100287896 LIPT2 PGM2L1 |
nsv555443 | Chr11:74398121 - 74469444 on Build GRCh38 | Loss | MIR548AL KCNE3 PGM2L1 |
nsv832209 | Chr11:74444838 - 74618074 on Build GRCh38 | Gain | POLD3 KCNE3 LOC100287896 LIPT2 |
dgv209e214 | Chr11:74225600 - 74828943 on Build GRCh38 | Gain | PPME1 MIR4696 P4HA3 CHRDL2 RNF169 MIR548AL POLD3 KCNE3 LOC101928580 LOC100287896 LIPT2 PGM2L1 |
nsv393 | Chr11:74424817 - 74470372 on Build GRCh38 | Deletion | KCNE3 |
nsv825994 | Chr11:74225622 - 74823410 on Build GRCh38 | Gain | PPME1 MIR4696 P4HA3 CHRDL2 RNF169 MIR548AL POLD3 KCNE3 LOC101928580 LOC100287896 LIPT2 PGM2L1 |