Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74454763-74455274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNE3
Entrez Gene ID
10008
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 3
Gene Aliases
HOKPP, HYPP, MiRP2
UniGene
Hs.523899
Gene Chromosome Location
Chr.11: 74454841-74467729 on Build GRCh38
Transcripts
NM_005472.4 XM_017017047.1 XM_017017048.1 XM_017017049.1 XM_017017050.1 XM_017017051.1 XM_017017052.1

SNP Detail

SNP ID Chromosome Location
rs72550286 Chr.11: 74455010-74455010 on Build GRCh38
rs927905659 Chr.11: 74455140-74455140 on Build GRCh38
rs920254635 Chr.11: 74454927-74454927 on Build GRCh38
rs1019214292 Chr.11: 74454972-74454972 on Build GRCh38
rs1045031211 Chr.11: 74454871-74454871 on Build GRCh38
rs904508209 Chr.11: 74455134-74455134 on Build GRCh38
rs541444340 Chr.11: 74455027-74455027 on Build GRCh38
rs947881161 Chr.11: 74455088-74455088 on Build GRCh38
rs142524202 Chr.11: 74455066-74455066 on Build GRCh38
rs906413355 Chr.11: 74454884-74454884 on Build GRCh38
rs939229324 Chr.11: 74455148-74455148 on Build GRCh38
rs181327394 Chr.11: 74455025-74455025 on Build GRCh38
rs986235282 Chr.11: 74454968-74454968 on Build GRCh38
rs751698690 Chr.11: 74455083-74455083 on Build GRCh38
rs779620464 Chr.11: 74455193-74455193 on Build GRCh38
rs1002006323 Chr.11: 74454837-74454837 on Build GRCh38
rs973100436 Chr.11: 74454920-74454920 on Build GRCh38
rs911409411 Chr.11: 74455011-74455011 on Build GRCh38
rs977458049 Chr.11: 74455004-74455004 on Build GRCh38
rs1043085611 Chr.11: 74455094-74455094 on Build GRCh38
rs1014062916 Chr.11: 74455209-74455209 on Build GRCh38
rs897533008 Chr.11: 74455152-74455152 on Build GRCh38
rs931776699 Chr.11: 74455008-74455008 on Build GRCh38
rs780233556 Chr.11: 74454984-74454984 on Build GRCh38
rs888843292 Chr.11: 74455203-74455203 on Build GRCh38
rs535500151 Chr.11: 74454912-74454912 on Build GRCh38
rs1022033944 Chr.11: 74454802-74454802 on Build GRCh38
rs1048812185 Chr.11: 74455186-74455186 on Build GRCh38
rs566273812 Chr.11: 74455044-74455044 on Build GRCh38
rs947852087 Chr.11: 74454852-74454852 on Build GRCh38
rs990258105 Chr.11: 74455072-74455072 on Build GRCh38
rs1054537586 Chr.11: 74454834-74454834 on Build GRCh38
rs961714523 Chr.11: 74454918-74454918 on Build GRCh38
rs1014757781 Chr.11: 74455213-74455213 on Build GRCh38
rs549819195 Chr.11: 74454899-74454899 on Build GRCh38
rs34315739 Chr.11: 74454909-74454909 on Build GRCh38
rs114922048 Chr.11: 74454812-74454812 on Build GRCh38
rs1027355548 Chr.11: 74454928-74454928 on Build GRCh38
rs188613083 Chr.11: 74454976-74454976 on Build GRCh38
rs1045277981 Chr.11: 74455106-74455106 on Build GRCh38
rs1034215014 Chr.11: 74454898-74454898 on Build GRCh38
rs777773780 Chr.11: 74455197-74455197 on Build GRCh38
rs966659193 Chr.11: 74454999-74454999 on Build GRCh38
rs913518578 Chr.11: 74455046-74455046 on Build GRCh38
rs886048645 Chr.11: 74455156-74455156 on Build GRCh38
rs1036271202 Chr.11: 74455150-74455150 on Build GRCh38
rs558085409 Chr.11: 74455112-74455112 on Build GRCh38
rs953578050 Chr.11: 74454957-74454957 on Build GRCh38
rs1019499496 Chr.11: 74455222-74455222 on Build GRCh38
rs755009687 Chr.11: 74455173-74455173 on Build GRCh38
rs1001638986 Chr.11: 74455169-74455169 on Build GRCh38
rs9516 Chr.11: 74455233-74455233 on Build GRCh38
rs992763277 Chr.11: 74454911-74454911 on Build GRCh38

View Full Product Details