Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74456380-74456924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNE3
Entrez Gene ID
10008
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 3
Gene Aliases
HOKPP, HYPP, MiRP2
UniGene
Hs.523899
Gene Chromosome Location
Chr.11: 74454841-74467729 on Build GRCh38
Transcripts
NM_005472.4 XM_011544713.2 XM_017017047.1 XM_017017048.1 XM_017017049.1 XM_017017050.1 XM_017017051.1 XM_017017052.1

SNP Detail

SNP ID Chromosome Location
rs1009154740 Chr.11: 74456443-74456443 on Build GRCh38
rs886048652 Chr.11: 74456516-74456516 on Build GRCh38
rs867068585 Chr.11: 74456894-74456894 on Build GRCh38
rs958875695 Chr.11: 74456492-74456492 on Build GRCh38
rs757178795 Chr.11: 74456675-74456675 on Build GRCh38
rs984561665 Chr.11: 74456780-74456780 on Build GRCh38
rs914309329 Chr.11: 74456839-74456839 on Build GRCh38
rs78749606 Chr.11: 74456690-74456690 on Build GRCh38
rs750800404 Chr.11: 74456889-74456889 on Build GRCh38
rs952254394 Chr.11: 74456645-74456645 on Build GRCh38
rs1033626876 Chr.11: 74456491-74456491 on Build GRCh38
rs1027009558 Chr.11: 74456685-74456685 on Build GRCh38
rs571939861 Chr.11: 74456448-74456448 on Build GRCh38
rs72552105 Chr.11: 74456485-74456485 on Build GRCh38
rs945294265 Chr.11: 74456824-74456824 on Build GRCh38
rs903640220 Chr.11: 74456467-74456467 on Build GRCh38
rs901517968 Chr.11: 74456419-74456419 on Build GRCh38
rs758082514 Chr.11: 74456795-74456795 on Build GRCh38
rs903505730 Chr.11: 74456873-74456873 on Build GRCh38
rs1001038148 Chr.11: 74456895-74456895 on Build GRCh38
rs12277342 Chr.11: 74456789-74456789 on Build GRCh38
rs886048653 Chr.11: 74456663-74456663 on Build GRCh38
rs72552107 Chr.11: 74456823-74456823 on Build GRCh38
rs919244531 Chr.11: 74456614-74456614 on Build GRCh38
rs542452253 Chr.11: 74456835-74456835 on Build GRCh38
rs572908442 Chr.11: 74456545-74456545 on Build GRCh38
rs951910298 Chr.11: 74456779-74456779 on Build GRCh38
rs1052848060 Chr.11: 74456451-74456451 on Build GRCh38
rs984863824 Chr.11: 74456650-74456650 on Build GRCh38
rs12271177 Chr.11: 74456585-74456585 on Build GRCh38
rs991760435 Chr.11: 74456542-74456542 on Build GRCh38
rs893346665 Chr.11: 74456453-74456453 on Build GRCh38
rs771985974 Chr.11: 74456886-74456886 on Build GRCh38
rs1020569695 Chr.11: 74456455-74456455 on Build GRCh38
rs190735627 Chr.11: 74456464-74456464 on Build GRCh38
rs41312921 Chr.11: 74456589-74456589 on Build GRCh38
rs557447799 Chr.11: 74456762-74456762 on Build GRCh38
rs1040275159 Chr.11: 74456794-74456794 on Build GRCh38
rs192619152 Chr.11: 74456610-74456610 on Build GRCh38
rs150406522 Chr.11: 74456848-74456848 on Build GRCh38
rs1003290138 Chr.11: 74456564-74456564 on Build GRCh38
rs182905404 Chr.11: 74456804-74456804 on Build GRCh38
rs906085047 Chr.11: 74456540-74456540 on Build GRCh38
rs543280723 Chr.11: 74456788-74456788 on Build GRCh38
rs534651370 Chr.11: 74456678-74456678 on Build GRCh38
rs988581888 Chr.11: 74456815-74456815 on Build GRCh38
rs72552106 Chr.11: 74456778-74456778 on Build GRCh38
rs41312919 Chr.11: 74456512-74456512 on Build GRCh38
rs934393281 Chr.11: 74456405-74456405 on Build GRCh38
rs187801915 Chr.11: 74456547-74456547 on Build GRCh38
rs947515764 Chr.11: 74456459-74456459 on Build GRCh38

View Full Product Details