Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102943219-102943753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP13
Entrez Gene ID
4322
Gene Name
matrix metallopeptidase 13
Gene Aliases
CLG3, MANDP1, MMP-13
UniGene
Hs.2936
Gene Chromosome Location
Chr.11: 102942992-102955734 on Build GRCh38
Transcripts
NM_002427.3

SNP Detail

SNP ID Chromosome Location
rs189328192 Chr.11: 102943466-102943466 on Build GRCh38
rs886047554 Chr.11: 102943510-102943510 on Build GRCh38
rs886047551 Chr.11: 102943259-102943259 on Build GRCh38
rs782048471 Chr.11: 102943371-102943371 on Build GRCh38
rs781904671 Chr.11: 102943251-102943252 on Build GRCh38
rs1043828307 Chr.11: 102943546-102943546 on Build GRCh38
rs938587284 Chr.11: 102943411-102943411 on Build GRCh38
rs534632404 Chr.11: 102943612-102943612 on Build GRCh38
rs782016341 Chr.11: 102943387-102943387 on Build GRCh38
rs891145205 Chr.11: 102943670-102943670 on Build GRCh38
rs373064452 Chr.11: 102943697-102943697 on Build GRCh38
rs1042840 Chr.11: 102943381-102943381 on Build GRCh38
rs114428333 Chr.11: 102943638-102943638 on Build GRCh38
rs536869199 Chr.11: 102943724-102943724 on Build GRCh38
rs1052425573 Chr.11: 102943639-102943639 on Build GRCh38
rs925360025 Chr.11: 102943634-102943634 on Build GRCh38
rs782162994 Chr.11: 102943600-102943600 on Build GRCh38
rs886047552 Chr.11: 102943416-102943417 on Build GRCh38
rs571076063 Chr.11: 102943666-102943666 on Build GRCh38
rs565834939 Chr.11: 102943549-102943549 on Build GRCh38
rs886047553 Chr.11: 102943495-102943495 on Build GRCh38
rs1008467700 Chr.11: 102943716-102943716 on Build GRCh38
rs770044566 Chr.11: 102943663-102943663 on Build GRCh38
rs532528718 Chr.11: 102943414-102943414 on Build GRCh38
rs35477433 Chr.11: 102943268-102943269 on Build GRCh38
rs915733663 Chr.11: 102943450-102943450 on Build GRCh38

View Full Product Details