Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113417944-113418429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD2
Entrez Gene ID
1813
Gene Name
dopamine receptor D2
Gene Aliases
D2DR, D2R
UniGene
Hs.73893
Gene Chromosome Location
Chr.11: 113409595-113475279 on Build GRCh38
Transcripts
NM_000795.3 NM_016574.3 XM_017017296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3985936 c.297G>A p.E99E Chr.11: 113418125-113418125 on Build GRCh38
4869447 c.302A>C p.K101T Chr.11: 113418120-113418120 on Build GRCh38
4599144 c.314T>C p.I105T Chr.11: 113418108-113418108 on Build GRCh38
3985935 c.297G>A p.E99E Chr.11: 113418125-113418125 on Build GRCh38
923196 c.302A>C p.K101T Chr.11: 113418120-113418120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2075654 Chr.11: 113418344-113418344 on Build GRCh38
rs148478679 Chr.11: 113418044-113418044 on Build GRCh38
rs773774927 Chr.11: 113418165-113418165 on Build GRCh38
rs547324394 Chr.11: 113418190-113418190 on Build GRCh38
rs766789195 Chr.11: 113418173-113418173 on Build GRCh38
rs1018087709 Chr.11: 113418365-113418365 on Build GRCh38
rs895288765 Chr.11: 113418286-113418286 on Build GRCh38
rs986889547 Chr.11: 113417972-113417972 on Build GRCh38
rs752361704 Chr.11: 113418053-113418053 on Build GRCh38
rs748688997 Chr.11: 113418163-113418169 on Build GRCh38
rs954248271 Chr.11: 113418252-113418252 on Build GRCh38
rs770900138 Chr.11: 113417979-113417979 on Build GRCh38
rs549053606 Chr.11: 113418031-113418031 on Build GRCh38
rs776974862 Chr.11: 113418295-113418295 on Build GRCh38
rs200503273 Chr.11: 113417985-113417985 on Build GRCh38
rs75646654 Chr.11: 113418390-113418390 on Build GRCh38
rs942255389 Chr.11: 113418259-113418259 on Build GRCh38
rs750801533 Chr.11: 113418032-113418032 on Build GRCh38
rs576575938 Chr.11: 113418257-113418257 on Build GRCh38
rs776612281 Chr.11: 113418144-113418144 on Build GRCh38
rs965562072 Chr.11: 113418389-113418389 on Build GRCh38
rs916448747 Chr.11: 113418353-113418353 on Build GRCh38
rs893759807 Chr.11: 113418333-113418333 on Build GRCh38
rs999205009 Chr.11: 113418208-113418208 on Build GRCh38
rs145674979 Chr.11: 113418216-113418216 on Build GRCh38
rs919907050 Chr.11: 113418314-113418314 on Build GRCh38
rs1046631577 Chr.11: 113418399-113418399 on Build GRCh38
rs1011763452 Chr.11: 113418227-113418227 on Build GRCh38
rs999444731 Chr.11: 113418310-113418310 on Build GRCh38
rs200402411 Chr.11: 113418080-113418080 on Build GRCh38
rs192602141 Chr.11: 113418228-113418228 on Build GRCh38
rs745749347 Chr.11: 113418091-113418091 on Build GRCh38
rs775364912 Chr.11: 113418116-113418116 on Build GRCh38
rs1031936808 Chr.11: 113418212-113418212 on Build GRCh38
rs776836586 Chr.11: 113418018-113418018 on Build GRCh38
rs976890303 Chr.11: 113418175-113418175 on Build GRCh38
rs199754592 Chr.11: 113418062-113418062 on Build GRCh38
rs550928971 Chr.11: 113418154-113418154 on Build GRCh38
rs745657838 Chr.11: 113417986-113417986 on Build GRCh38
rs547664686 Chr.11: 113418152-113418152 on Build GRCh38
rs200747382 Chr.11: 113418184-113418184 on Build GRCh38
rs1055332966 Chr.11: 113418271-113418271 on Build GRCh38
rs931210463 Chr.11: 113418034-113418034 on Build GRCh38
rs541708544 Chr.11: 113418359-113418359 on Build GRCh38
rs902521282 Chr.11: 113418309-113418309 on Build GRCh38
rs983710550 Chr.11: 113418217-113418217 on Build GRCh38
rs200429548 Chr.11: 113418008-113418008 on Build GRCh38
rs377113965 Chr.11: 113418151-113418151 on Build GRCh38
rs965186890 Chr.11: 113418171-113418171 on Build GRCh38
rs754957378 Chr.11: 113418157-113418157 on Build GRCh38
rs372879445 Chr.11: 113418040-113418040 on Build GRCh38
rs768995013 Chr.11: 113418135-113418135 on Build GRCh38
rs902271274 Chr.11: 113418401-113418401 on Build GRCh38
rs200713577 Chr.11: 113418159-113418159 on Build GRCh38
rs200906213 Chr.11: 113417991-113417991 on Build GRCh38
rs201130366 Chr.11: 113418092-113418092 on Build GRCh38
rs764280522 Chr.11: 113418014-113418014 on Build GRCh38
rs1107162 Chr.11: 113418315-113418315 on Build GRCh38
rs748803426 Chr.11: 113418068-113418068 on Build GRCh38
rs199765712 Chr.11: 113418127-113418127 on Build GRCh38
rs762292044 Chr.11: 113418024-113418024 on Build GRCh38
rs778935653 Chr.11: 113418086-113418086 on Build GRCh38
rs200846690 Chr.11: 113418182-113418182 on Build GRCh38
rs749245915 Chr.11: 113417976-113417976 on Build GRCh38
rs201724929 Chr.11: 113418063-113418063 on Build GRCh38
rs536377918 Chr.11: 113418218-113418218 on Build GRCh38
rs747973563 Chr.11: 113418207-113418207 on Build GRCh38
rs202075929 Chr.11: 113418003-113418003 on Build GRCh38
rs765191561 Chr.11: 113418160-113418160 on Build GRCh38
rs146581145 Chr.11: 113418293-113418293 on Build GRCh38
rs561364046 Chr.11: 113418386-113418386 on Build GRCh38
rs141811103 Chr.11: 113418065-113418065 on Build GRCh38
rs571218398 Chr.11: 113418167-113418167 on Build GRCh38
rs912667210 Chr.11: 113418270-113418270 on Build GRCh38
rs201393591 Chr.11: 113418177-113418177 on Build GRCh38

View Full Product Details