Product Details

Assay Reference Genome
Location

Chr.6:161349827 on build GRCh38
Cytoband
6q26
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
5071
Gene Symbol
PARK2
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
Location
Chr.6:161347417-162727802 on build GRCh38
Assay Gene Location
Within Exon 13
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PARK2 NM_004562.2 12 1804 NP_004553.2
NM_013987.2 11 1720 NP_054642.2
NM_013988.2 9 1357 NP_054643.2
XM_011535863.1 12 1801 XP_011534165.1
XM_017010908.1 12 2718 XP_016866397.1
XM_017010909.1 11 1598 XP_016866398.1
AB009973.1 12 1771 BAA25751.1
AK294684.1 7 1010

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv830854 Chr6:161318894 - 161506483 on Build GRCh38 Gain PARK2 LOC105378098
nsv1029727 Chr6:161186648 - 161393263 on Build GRCh38 Gain AGPAT4 PARK2
nsv1031234 Chr6:161255736 - 161829694 on Build GRCh38 Gain AGPAT4 PARK2 LOC105378098
esv3576244 Chr6:161272053 - 161395540 on Build GRCh38 Gain AGPAT4 PARK2
nsv1026543 Chr6:160306836 - 161585804 on Build GRCh38 Gain PLG AGPAT4 PARK2 AGPAT4-IT1 LPA LPAL2 LOC105378098 SLC22A3 MAP3K4
esv3576243 Chr6:161178374 - 161396606 on Build GRCh38 Gain AGPAT4 PARK2
esv2752091 Chr6:161171068 - 161434753 on Build GRCh38 Gain AGPAT4 PARK2

View Full Product Details