Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
KCNE3 | NM_005472.4 | 3 | 2697 | NP_005463.1 |
XM_017017047.1 | 4 | 2767 | XP_016872536.1 | |
XM_017017048.1 | 5 | 2863 | XP_016872537.1 | |
XM_017017049.1 | 3 | 2713 | XP_016872538.1 | |
XM_017017050.1 | 3 | 5571 | XP_016872539.1 | |
XM_017017051.1 | 4 | 2686 | XP_016872540.1 | |
XM_017017052.1 | 3 | 2637 | XP_016872541.1 | |
BC113743.1 | 2 | 2521 | AAI13744.1 | |
CK301015.1 | 1 | 390 | ||
CR936625.1 | 4 | 2487 | CAI56768.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1041154 | Chr11:74403756 - 74466864 on Build GRCh38 | Loss | KCNE3 |
esv34979 | Chr11:74167643 - 74822407 on Build GRCh38 | Gain | LOC100287896 LIPT2 LOC101928580 POLD3 MIR4696 RNF169 P4HA3 CHRDL2 PPME1 C2CD3 KCNE3 MIR548AL PGM2L1 |
esv3626840 | Chr11:74371082 - 74464389 on Build GRCh38 | Gain | KCNE3 MIR548AL PGM2L1 |
esv2759837 | Chr11:74131564 - 74846552 on Build GRCh38 | Gain | LOC100287896 XRRA1 LIPT2 LOC101928580 POLD3 MIR4696 RNF169 P4HA3 CHRDL2 PPME1 C2CD3 KCNE3 MIR548AL PGM2L1 |
nsv555443 | Chr11:74398121 - 74469444 on Build GRCh38 | Loss | KCNE3 MIR548AL PGM2L1 |
nsv832209 | Chr11:74444838 - 74618074 on Build GRCh38 | Gain | LOC100287896 LIPT2 KCNE3 POLD3 |
dgv209e214 | Chr11:74225600 - 74828943 on Build GRCh38 | Gain | RNF169 P4HA3 CHRDL2 LOC100287896 PPME1 LIPT2 KCNE3 MIR548AL LOC101928580 PGM2L1 POLD3 MIR4696 |
nsv393 | Chr11:74424817 - 74470372 on Build GRCh38 | Deletion | KCNE3 |
nsv825994 | Chr11:74225622 - 74823410 on Build GRCh38 | Gain | RNF169 P4HA3 CHRDL2 LOC100287896 PPME1 LIPT2 KCNE3 MIR548AL LOC101928580 PGM2L1 POLD3 MIR4696 |