Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20187759-20188308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJB2
Entrez Gene ID
2706
Gene Name
gap junction protein beta 2
Gene Aliases
CX26, DFNA3, DFNA3A, DFNB1, DFNB1A, HID, KID, NSRD1, PPK
UniGene
Hs.524894
Gene Chromosome Location
Chr.13: 20187463-20192975 on Build GRCh38
Transcripts
NM_004004.5 XM_011535049.2

SNP Detail

SNP ID Chromosome Location
rs1016943512 Chr.13: 20188265-20188265 on Build GRCh38
rs147595498 Chr.13: 20188176-20188176 on Build GRCh38
rs1026971249 Chr.13: 20187943-20187943 on Build GRCh38
rs1047934427 Chr.13: 20188195-20188195 on Build GRCh38
rs35136382 Chr.13: 20188008-20188009 on Build GRCh38
rs966180251 Chr.13: 20188259-20188259 on Build GRCh38
rs895120248 Chr.13: 20188273-20188273 on Build GRCh38
rs181728208 Chr.13: 20188058-20188058 on Build GRCh38
rs936634374 Chr.13: 20188083-20188083 on Build GRCh38
rs1004577569 Chr.13: 20188208-20188208 on Build GRCh38
rs1006432105 Chr.13: 20188222-20188222 on Build GRCh38
rs9237 Chr.13: 20187834-20187834 on Build GRCh38
rs764456740 Chr.13: 20187869-20187869 on Build GRCh38
rs757704123 Chr.13: 20187961-20187961 on Build GRCh38
rs989812267 Chr.13: 20188098-20188098 on Build GRCh38
rs959366452 Chr.13: 20187988-20187988 on Build GRCh38
rs886685299 Chr.13: 20188196-20188196 on Build GRCh38
rs537683957 Chr.13: 20187885-20187885 on Build GRCh38
rs149900249 Chr.13: 20187979-20187979 on Build GRCh38
rs187158699 Chr.13: 20188115-20188115 on Build GRCh38
rs5030700 Chr.13: 20187970-20187970 on Build GRCh38
rs910849327 Chr.13: 20188101-20188101 on Build GRCh38
rs1019095526 Chr.13: 20188239-20188239 on Build GRCh38
rs886050027 Chr.13: 20188101-20188101 on Build GRCh38
rs933458665 Chr.13: 20188175-20188175 on Build GRCh38
rs866743521 Chr.13: 20188126-20188126 on Build GRCh38
rs185790172 Chr.13: 20187868-20187868 on Build GRCh38
rs990838225 Chr.13: 20187997-20187997 on Build GRCh38
rs762265299 Chr.13: 20187969-20187973 on Build GRCh38
rs559171604 Chr.13: 20187969-20187969 on Build GRCh38
rs111729919 Chr.13: 20187916-20187916 on Build GRCh38
rs111732260 Chr.13: 20188143-20188143 on Build GRCh38
rs752926267 Chr.13: 20187819-20187819 on Build GRCh38
rs886204715 Chr.13: 20188200-20188200 on Build GRCh38
rs746404073 Chr.13: 20188052-20188052 on Build GRCh38
rs997698706 Chr.13: 20187937-20187937 on Build GRCh38
rs923851905 Chr.13: 20188079-20188079 on Build GRCh38
rs901989526 Chr.13: 20187890-20187890 on Build GRCh38
rs867312300 Chr.13: 20188104-20188104 on Build GRCh38
rs1025026812 Chr.13: 20188054-20188054 on Build GRCh38
rs535474422 Chr.13: 20188033-20188033 on Build GRCh38
rs546826225 Chr.13: 20187922-20187922 on Build GRCh38
rs912033122 Chr.13: 20187861-20187861 on Build GRCh38
rs189685514 Chr.13: 20187986-20187986 on Build GRCh38
rs977974907 Chr.13: 20188076-20188076 on Build GRCh38
rs535822487 Chr.13: 20187814-20187814 on Build GRCh38

View Full Product Details