Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12193835-12194372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF1B
Entrez Gene ID
7133
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
UniGene
Hs.256278
Gene Chromosome Location
Chr.1: 12166948-12209222 on Build GRCh38
Transcripts
NM_001066.2 XM_011542060.2 XM_011542063.2 XM_017002211.1 XM_017002212.1 XM_017002213.1 XM_017002214.1 XM_017002215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7111663 c.813G>C p.L271F Chr.1: 12193980-12193980 on Build GRCh38
3417963 c.788-1G>T p.? Chr.1: 12193954-12193954 on Build GRCh38
6533573 c.826A>G p.I276V Chr.1: 12193993-12193993 on Build GRCh38
5908034 c.788-4C>T p.? Chr.1: 12193951-12193951 on Build GRCh38
4853201 c.849C>T p.I283I Chr.1: 12194016-12194016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372488130 Chr.1: 12193862-12193862 on Build GRCh38
rs951619189 Chr.1: 12193961-12193961 on Build GRCh38
rs748589747 Chr.1: 12193960-12193960 on Build GRCh38
rs201648206 Chr.1: 12194022-12194022 on Build GRCh38
rs1017742507 Chr.1: 12193872-12193872 on Build GRCh38
rs113160184 Chr.1: 12194033-12194033 on Build GRCh38
rs758369350 Chr.1: 12194036-12194036 on Build GRCh38
rs775210038 Chr.1: 12193970-12193970 on Build GRCh38
rs934021492 Chr.1: 12194097-12194097 on Build GRCh38
rs570659072 Chr.1: 12193918-12193918 on Build GRCh38
rs187804577 Chr.1: 12193864-12193864 on Build GRCh38
rs1019971326 Chr.1: 12194328-12194328 on Build GRCh38
rs926662964 Chr.1: 12194111-12194111 on Build GRCh38
rs557988205 Chr.1: 12194285-12194285 on Build GRCh38
rs765198273 Chr.1: 12194260-12194260 on Build GRCh38
rs980147628 Chr.1: 12193863-12193863 on Build GRCh38
rs563979080 Chr.1: 12193994-12193994 on Build GRCh38
rs945254812 Chr.1: 12194209-12194209 on Build GRCh38
rs1004706588 Chr.1: 12194124-12194124 on Build GRCh38
rs960018818 Chr.1: 12193915-12193917 on Build GRCh38
rs769055591 Chr.1: 12193955-12193955 on Build GRCh38
rs1043641200 Chr.1: 12194212-12194212 on Build GRCh38
rs61761336 Chr.1: 12193987-12193987 on Build GRCh38
rs998269059 Chr.1: 12193925-12193925 on Build GRCh38
rs781422111 Chr.1: 12193861-12193861 on Build GRCh38
rs763557181 Chr.1: 12194044-12194047 on Build GRCh38
rs143074721 Chr.1: 12194283-12194283 on Build GRCh38
rs998339444 Chr.1: 12194284-12194284 on Build GRCh38
rs760348211 Chr.1: 12194121-12194121 on Build GRCh38
rs539270101 Chr.1: 12193922-12193922 on Build GRCh38
rs903531635 Chr.1: 12194243-12194243 on Build GRCh38
rs761885846 Chr.1: 12194202-12194202 on Build GRCh38
rs375195181 Chr.1: 12194089-12194089 on Build GRCh38
rs887215574 Chr.1: 12194123-12194123 on Build GRCh38
rs776244396 Chr.1: 12194159-12194159 on Build GRCh38
rs530563938 Chr.1: 12193880-12193880 on Build GRCh38
rs547792261 Chr.1: 12193858-12193858 on Build GRCh38
rs375449058 Chr.1: 12194035-12194035 on Build GRCh38
rs749686651 Chr.1: 12193936-12193936 on Build GRCh38
rs776907951 Chr.1: 12194002-12194002 on Build GRCh38
rs1007603256 Chr.1: 12194245-12194245 on Build GRCh38
rs780871115 Chr.1: 12193927-12193927 on Build GRCh38
rs17879042 Chr.1: 12193972-12193972 on Build GRCh38
rs111772337 Chr.1: 12194174-12194174 on Build GRCh38
rs763716271 Chr.1: 12194041-12194041 on Build GRCh38
rs760722962 Chr.1: 12193980-12193980 on Build GRCh38
rs201668794 Chr.1: 12193908-12193908 on Build GRCh38
rs946215007 Chr.1: 12193982-12193982 on Build GRCh38
rs959724871 Chr.1: 12194277-12194277 on Build GRCh38
rs756791462 Chr.1: 12194060-12194060 on Build GRCh38
rs757993456 Chr.1: 12193904-12193904 on Build GRCh38
rs2275416 Chr.1: 12194144-12194144 on Build GRCh38
rs1001461917 Chr.1: 12194214-12194214 on Build GRCh38
rs535844061 Chr.1: 12194057-12194057 on Build GRCh38
rs2229700 Chr.1: 12193958-12193958 on Build GRCh38
rs375458572 Chr.1: 12194172-12194172 on Build GRCh38
rs780770003 Chr.1: 12194061-12194061 on Build GRCh38
rs192393562 Chr.1: 12194026-12194026 on Build GRCh38
rs376618055 Chr.1: 12194303-12194303 on Build GRCh38
rs779671137 Chr.1: 12194142-12194142 on Build GRCh38
rs773229323 Chr.1: 12193976-12193976 on Build GRCh38

View Full Product Details