Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 31276615-31277108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GLCT
Entrez Gene ID
145173
Gene Name
beta 3-glucosyltransferase
Gene Aliases
B3GALTL, B3GTL, B3Glc-T, Gal-T, beta3Glc-T
UniGene
Hs.13205
Gene Chromosome Location
Chr.13: 31199975-31332276 on Build GRCh38
Transcripts
NM_194318.3 XM_006719768.3 XM_011534936.1 XM_011534938.2 XM_017020395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
238929 c.787C>T p.P263S Chr.13: 31276708-31276708 on Build GRCh38
6609256 c.819delA p.T275fs*61 Chr.13: 31276740-31276740 on Build GRCh38
946678 c.836T>G p.F279C Chr.13: 31276757-31276757 on Build GRCh38
7267383 c.781-10delT p.? Chr.13: 31276692-31276692 on Build GRCh38
3468294 c.838C>T p.H280Y Chr.13: 31276759-31276759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939901475 Chr.13: 31276637-31276637 on Build GRCh38
rs1009776912 Chr.13: 31276838-31276838 on Build GRCh38
rs781291553 Chr.13: 31276691-31276692 on Build GRCh38
rs373860865 Chr.13: 31276707-31276707 on Build GRCh38
rs773268537 Chr.13: 31276724-31276724 on Build GRCh38
rs759565970 Chr.13: 31276658-31276658 on Build GRCh38
rs770217847 Chr.13: 31276813-31276813 on Build GRCh38
rs775867735 Chr.13: 31276817-31276817 on Build GRCh38
rs887893330 Chr.13: 31276992-31276992 on Build GRCh38
rs769064477 Chr.13: 31276692-31276692 on Build GRCh38
rs766246764 Chr.13: 31276744-31276744 on Build GRCh38
rs191574496 Chr.13: 31276929-31276929 on Build GRCh38
rs1007637830 Chr.13: 31276993-31276993 on Build GRCh38
rs901305703 Chr.13: 31276884-31276884 on Build GRCh38
rs777619322 Chr.13: 31276899-31276899 on Build GRCh38
rs570386340 Chr.13: 31277062-31277062 on Build GRCh38
rs778270237 Chr.13: 31276659-31276659 on Build GRCh38
rs1048864586 Chr.13: 31276981-31276981 on Build GRCh38
rs754730230 Chr.13: 31276775-31276775 on Build GRCh38
rs1017593013 Chr.13: 31276720-31276720 on Build GRCh38
rs777256172 Chr.13: 31276796-31276796 on Build GRCh38
rs1055901578 Chr.13: 31276849-31276849 on Build GRCh38
rs551771976 Chr.13: 31277012-31277012 on Build GRCh38
rs966035607 Chr.13: 31277083-31277083 on Build GRCh38
rs950012362 Chr.13: 31276882-31276882 on Build GRCh38
rs775814597 Chr.13: 31276700-31276700 on Build GRCh38
rs375308431 Chr.13: 31276765-31276765 on Build GRCh38
rs747597532 Chr.13: 31276687-31276687 on Build GRCh38
rs376456091 Chr.13: 31276675-31276675 on Build GRCh38
rs1015547911 Chr.13: 31276957-31276957 on Build GRCh38
rs746247747 Chr.13: 31276802-31276802 on Build GRCh38
rs984517843 Chr.13: 31276953-31276953 on Build GRCh38
rs137981677 Chr.13: 31276666-31276667 on Build GRCh38
rs754211193 Chr.13: 31276969-31276969 on Build GRCh38
rs376790604 Chr.13: 31276977-31276977 on Build GRCh38
rs746150246 Chr.13: 31276677-31276677 on Build GRCh38
rs375057499 Chr.13: 31277011-31277012 on Build GRCh38
rs966731341 Chr.13: 31276723-31276723 on Build GRCh38
rs372388899 Chr.13: 31276801-31276801 on Build GRCh38
rs756035046 Chr.13: 31276883-31276883 on Build GRCh38
rs199615610 Chr.13: 31276782-31276782 on Build GRCh38
rs4065552 Chr.13: 31276852-31276852 on Build GRCh38
rs141291628 Chr.13: 31277009-31277010 on Build GRCh38
rs747589352 Chr.13: 31276797-31276797 on Build GRCh38
rs757958058 Chr.13: 31276789-31276789 on Build GRCh38
rs760500105 Chr.13: 31276735-31276735 on Build GRCh38
rs764256479 Chr.13: 31276709-31276709 on Build GRCh38
rs747454787 Chr.13: 31276672-31276672 on Build GRCh38
rs952066927 Chr.13: 31276668-31276668 on Build GRCh38
rs914062573 Chr.13: 31276759-31276759 on Build GRCh38
rs917037385 Chr.13: 31276666-31276673 on Build GRCh38
rs752292217 Chr.13: 31276788-31276788 on Build GRCh38
rs776827046 Chr.13: 31276676-31276676 on Build GRCh38
rs993550796 Chr.13: 31276642-31276642 on Build GRCh38
rs281865463 Chr.13: 31276671-31276672 on Build GRCh38
rs753600057 Chr.13: 31276752-31276752 on Build GRCh38
rs554800057 Chr.13: 31276917-31276917 on Build GRCh38
rs1409373 Chr.13: 31276922-31276922 on Build GRCh38
rs770160902 Chr.13: 31276692-31276692 on Build GRCh38
rs780390095 Chr.13: 31276679-31276680 on Build GRCh38
rs750915448 Chr.13: 31276998-31276998 on Build GRCh38

View Full Product Details