Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2602739-2603272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDPK1
Entrez Gene ID
5170
Gene Name
3-phosphoinositide dependent protein kinase 1
Gene Aliases
PDK1, PDPK2, PDPK2P, PRO0461
UniGene
Hs.459691
Gene Chromosome Location
Chr.16: 2537964-2603190 on Build GRCh38
Transcripts
NM_001261816.1 NM_002613.4 NM_031268.5 XM_011522521.2 XM_011522523.2 XM_017023271.1 XM_017023272.1 XM_017023273.1 XM_017023274.1 XM_017023275.1 XM_017023276.1 XM_017023277.1

SNP Detail

SNP ID Chromosome Location
rs1031130094 Chr.16: 2603226-2603226 on Build GRCh38
rs906520793 Chr.16: 2603148-2603148 on Build GRCh38
rs574208711 Chr.16: 2602852-2602852 on Build GRCh38
rs978306054 Chr.16: 2602800-2602800 on Build GRCh38
rs989643839 Chr.16: 2603079-2603080 on Build GRCh38
rs557863257 Chr.16: 2603117-2603117 on Build GRCh38
rs556860773 Chr.16: 2602939-2602939 on Build GRCh38
rs111618468 Chr.16: 2603088-2603089 on Build GRCh38
rs548902594 Chr.16: 2603232-2603232 on Build GRCh38
rs545626630 Chr.16: 2602848-2602848 on Build GRCh38
rs1025380467 Chr.16: 2603010-2603010 on Build GRCh38
rs1031180855 Chr.16: 2603064-2603064 on Build GRCh38
rs1016176078 Chr.16: 2602966-2602966 on Build GRCh38
rs922966530 Chr.16: 2603252-2603252 on Build GRCh38
rs11554592 Chr.16: 2603090-2603090 on Build GRCh38
rs866924043 Chr.16: 2603099-2603099 on Build GRCh38
rs1024986190 Chr.16: 2603218-2603218 on Build GRCh38
rs542840852 Chr.16: 2602868-2602868 on Build GRCh38
rs201869822 Chr.16: 2602890-2602891 on Build GRCh38
rs772425944 Chr.16: 2603070-2603070 on Build GRCh38
rs553056589 Chr.16: 2602879-2602879 on Build GRCh38
rs189105574 Chr.16: 2603143-2603143 on Build GRCh38
rs1047571085 Chr.16: 2602932-2602932 on Build GRCh38
rs933042097 Chr.16: 2602893-2602893 on Build GRCh38
rs533610862 Chr.16: 2602822-2602822 on Build GRCh38
rs1006097939 Chr.16: 2602947-2602947 on Build GRCh38
rs895059864 Chr.16: 2602991-2602991 on Build GRCh38
rs918044521 Chr.16: 2602785-2602785 on Build GRCh38
rs1039100683 Chr.16: 2602889-2602889 on Build GRCh38
rs941904765 Chr.16: 2603201-2603201 on Build GRCh38
rs970853219 Chr.16: 2603047-2603047 on Build GRCh38
rs1012552437 Chr.16: 2603208-2603208 on Build GRCh38
rs942933495 Chr.16: 2603094-2603094 on Build GRCh38
rs554095762 Chr.16: 2602832-2602832 on Build GRCh38
rs911509961 Chr.16: 2603089-2603089 on Build GRCh38
rs181213412 Chr.16: 2603095-2603095 on Build GRCh38
rs541541489 Chr.16: 2603151-2603151 on Build GRCh38
rs536544675 Chr.16: 2603246-2603246 on Build GRCh38
rs989676533 Chr.16: 2603237-2603237 on Build GRCh38
rs557285323 Chr.16: 2602796-2602796 on Build GRCh38
rs1018434960 Chr.16: 2603242-2603242 on Build GRCh38
rs576761849 Chr.16: 2603087-2603087 on Build GRCh38
rs568815093 Chr.16: 2603247-2603247 on Build GRCh38
rs879212326 Chr.16: 2603185-2603185 on Build GRCh38
rs11554591 Chr.16: 2603019-2603019 on Build GRCh38
rs540470378 Chr.16: 2602869-2602871 on Build GRCh38
rs571020949 Chr.16: 2602816-2602816 on Build GRCh38
rs776922049 Chr.16: 2603227-2603227 on Build GRCh38
rs143376348 Chr.16: 2603223-2603223 on Build GRCh38
rs527809139 Chr.16: 2603106-2603106 on Build GRCh38
rs185318792 Chr.16: 2603119-2603119 on Build GRCh38
rs748736402 Chr.16: 2602905-2602905 on Build GRCh38
rs906491836 Chr.16: 2603220-2603220 on Build GRCh38
rs958100128 Chr.16: 2603231-2603231 on Build GRCh38
rs924081462 Chr.16: 2602810-2602810 on Build GRCh38
rs971208284 Chr.16: 2602795-2602795 on Build GRCh38
rs1047896262 Chr.16: 2603107-2603107 on Build GRCh38
rs1012280702 Chr.16: 2602999-2603000 on Build GRCh38
rs978549916 Chr.16: 2603063-2603063 on Build GRCh38
rs557177623 Chr.16: 2603202-2603202 on Build GRCh38
rs201698119 Chr.16: 2603099-2603100 on Build GRCh38

View Full Product Details