The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001231.4 | NP_001222.3 | 6 - 7 | 1041 | 81 |
GenBank mRNA | AK300713.1 | - | 5 - 6 | 845 | 81 |
AK313250.1 | - | 6 - 7 | 944 | 81 | |
BC022289.1 | - | 6 - 7 | 927 | 81 | |
S73775.1 | - | 6 - 7 | 931 | 81 |