Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46674165-46674662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS35
Entrez Gene ID
55737
Gene Name
VPS35 retromer complex component
Gene Aliases
MEM3, PARK17
UniGene
Hs.454528
Gene Chromosome Location
Chr.16: 46657979-46689232 on Build GRCh38
Transcripts
NM_018206.5 XM_005256045.3 XM_011523227.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6820562 c.977A>G p.D326G Chr.16: 46674598-46674598 on Build GRCh38
3932356 c.948T>A p.P316P Chr.16: 46674627-46674627 on Build GRCh38
7309317 c.915-3_915-2insT p.? Chr.16: 46674662-46674663 on Build GRCh38
4260335 c.1094G>A p.R365H Chr.16: 46674380-46674380 on Build GRCh38
3888529 c.976G>A p.D326N Chr.16: 46674599-46674599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773750047 Chr.16: 46674468-46674468 on Build GRCh38
rs116254156 Chr.16: 46674329-46674329 on Build GRCh38
rs750731767 Chr.16: 46674511-46674511 on Build GRCh38
rs766573416 Chr.16: 46674538-46674538 on Build GRCh38
rs751508275 Chr.16: 46674217-46674217 on Build GRCh38
rs767613694 Chr.16: 46674587-46674587 on Build GRCh38
rs539934175 Chr.16: 46674281-46674281 on Build GRCh38
rs754226798 Chr.16: 46674336-46674336 on Build GRCh38
rs756040992 Chr.16: 46674597-46674597 on Build GRCh38
rs777437956 Chr.16: 46674602-46674602 on Build GRCh38
rs770226984 Chr.16: 46674628-46674628 on Build GRCh38
rs753216277 Chr.16: 46674548-46674548 on Build GRCh38
rs745416544 Chr.16: 46674615-46674615 on Build GRCh38
rs536411044 Chr.16: 46674270-46674270 on Build GRCh38
rs772970332 Chr.16: 46674322-46674322 on Build GRCh38
rs867293728 Chr.16: 46674446-46674446 on Build GRCh38
rs372739478 Chr.16: 46674504-46674504 on Build GRCh38
rs188883365 Chr.16: 46674454-46674454 on Build GRCh38
rs769692526 Chr.16: 46674418-46674418 on Build GRCh38
rs748655024 Chr.16: 46674627-46674627 on Build GRCh38
rs774611405 Chr.16: 46674632-46674632 on Build GRCh38
rs770029606 Chr.16: 46674629-46674629 on Build GRCh38
rs762131827 Chr.16: 46674350-46674350 on Build GRCh38
rs758568605 Chr.16: 46674381-46674381 on Build GRCh38
rs201593831 Chr.16: 46674380-46674380 on Build GRCh38
rs755079634 Chr.16: 46674557-46674557 on Build GRCh38
rs747944333 Chr.16: 46674616-46674616 on Build GRCh38
rs911222425 Chr.16: 46674354-46674354 on Build GRCh38
rs770335442 Chr.16: 46674326-46674326 on Build GRCh38
rs751599316 Chr.16: 46674393-46674393 on Build GRCh38
rs777098551 Chr.16: 46674488-46674488 on Build GRCh38
rs762573165 Chr.16: 46674323-46674323 on Build GRCh38
rs763097703 Chr.16: 46674526-46674526 on Build GRCh38
rs144346159 Chr.16: 46674630-46674630 on Build GRCh38
rs563975281 Chr.16: 46674439-46674439 on Build GRCh38
rs765489808 Chr.16: 46674355-46674355 on Build GRCh38
rs554451334 Chr.16: 46674273-46674273 on Build GRCh38
rs759041343 Chr.16: 46674483-46674483 on Build GRCh38
rs752668855 Chr.16: 46674596-46674596 on Build GRCh38
rs754892097 Chr.16: 46674394-46674394 on Build GRCh38
rs370809668 Chr.16: 46674502-46674502 on Build GRCh38
rs140265194 Chr.16: 46674432-46674432 on Build GRCh38
rs769188080 Chr.16: 46674485-46674485 on Build GRCh38
rs778590455 Chr.16: 46674613-46674613 on Build GRCh38
rs777184680 Chr.16: 46674620-46674620 on Build GRCh38
rs781234198 Chr.16: 46674399-46674399 on Build GRCh38
rs184427104 Chr.16: 46674278-46674278 on Build GRCh38
rs561467128 Chr.16: 46674517-46674517 on Build GRCh38
rs751620415 Chr.16: 46674550-46674550 on Build GRCh38
rs780001713 Chr.16: 46674387-46674387 on Build GRCh38
rs749058909 Chr.16: 46674609-46674609 on Build GRCh38
rs1014802303 Chr.16: 46674258-46674258 on Build GRCh38
rs564676491 Chr.16: 46674304-46674304 on Build GRCh38
rs756884549 Chr.16: 46674612-46674612 on Build GRCh38
rs902736653 Chr.16: 46674445-46674445 on Build GRCh38
rs747997807 Chr.16: 46674415-46674415 on Build GRCh38
rs113154062 Chr.16: 46674364-46674364 on Build GRCh38
rs780094120 Chr.16: 46674263-46674263 on Build GRCh38
rs754978683 Chr.16: 46674332-46674332 on Build GRCh38
rs200578690 Chr.16: 46674636-46674636 on Build GRCh38

View Full Product Details