The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001549.5 | NP_001540.2 | 1 - 2 | 267 | 70 |
GenBank mRNA | AF083470.1 | - | 1 - 2 | 90 | 70 |
AK290427.1 | - | 1 - 2 | 177 | 70 | |
AK312675.1 | - | 1 - 2 | 87 | 70 | |
DA081546.1 | - | 1 - 2 | 241 | 70 | |
DA160435.1 | - | 1 - 2 | 271 | 70 | |
U52513.1 | - | 1 - 2 | 73 | 70 |