Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GFAP
Entrez Gene ID
2670
Gene Name
glial fibrillary acidic protein
Gene Aliases
ALXDRD
Chromosome Location
Chr.17: 44905626 - 44915552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002055.4 NP_002046.1 7 - 8 1235 75
GenBank mRNA AF419299.1 - 7 - 8 1174 75
AK098380.1 - 8 - 9 1121 75
AK098758.1 - 5 - 6 874 75
AK222683.1 - 7 - 8 1183 75
AK296259.1 - 8 - 9 1132 75
AK303728.1 - 8 - 9 1108 75
AK315398.1 - 7 - 8 1189 75
BC013596.1 - 7 - 8 1171 75
BC041765.1 - 7 - 8 1175 75
BC062609.1 - 7 - 8 1168 75
BC127871.1 - 2 - 3 250 75
J04569.1 - 7 - 8 1172 75
M26638.1 - 2 - 3 117 75
S40719.1 - 7 - 8 1183 75

View Full Product Details