The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002237.3 | NP_002228.2 | 2 - 3 | 1074 | 97 |
XM_006723785.2 | XP_006723848.1 | 4 - 5 | 1457 | 97 | |
XM_011528800.1 | XP_011527102.1 | 3 - 4 | 1132 | 97 | |
XM_011528801.1 | XP_011527103.1 | 4 - 5 | 1241 | 97 | |
XM_011528802.1 | XP_011527104.1 | 4 - 5 | 1236 | 97 | |
XM_011528803.2 | XP_011527105.1 | 4 - 5 | 1163 | 97 | |
XM_011528804.1 | XP_011527106.1 | 3 - 4 | 1386 | 97 | |
XM_011528805.2 | XP_011527107.1 | 2 - 3 | 3297 | 97 | |
XM_011528806.1 | XP_011527108.1 | 2 - 3 | 1278 | 97 | |
GenBank mRNA | AF033383.1 | - | 1 - 2 | 779 | 97 |
AK290689.1 | - | 2 - 3 | 1075 | 97 | |
BC046629.2 | - | 2 - 3 | 1043 | 97 |