The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204171.1 | NP_001191100.1 | 3 - 4 | 322 | 88 |
NM_001278516.1 | NP_001265445.1 | 3 - 4 | 322 | 88 | |
NM_001278518.1 | NP_001265447.1 | 3 - 4 | 322 | 88 | |
NM_002393.4 | NP_002384.2 | 3 - 4 | 322 | 88 | |
XM_017001311.1 | XP_016856800.1 | 3 - 4 | 321 | 88 | |
XM_017001312.1 | XP_016856801.1 | 3 - 4 | 321 | 88 | |
XM_017001313.1 | XP_016856802.1 | 3 - 4 | 321 | 88 | |
GenBank mRNA | AF007111.1 | - | 3 - 4 | 272 | 88 |
AK223228.1 | - | 3 - 4 | 285 | 88 | |
AK303626.1 | - | 3 - 4 | 295 | 88 | |
AK304698.1 | - | 3 - 4 | 295 | 88 | |
AK310607.1 | - | 3 - 4 | 298 | 88 | |
AK310903.1 | - | 3 - 4 | 295 | 88 | |
AY949028.1 | - | 2 - 3 | 156 | 88 | |
BC067299.1 | - | 3 - 4 | 312 | 88 | |
BC105106.1 | - | 3 - 4 | 215 | 88 | |
BC143431.1 | - | 3 - 4 | 215 | 88 | |
BC143432.1 | - | 3 - 4 | 215 | 88 | |
BP247507.1 | - | 3 - 4 | 322 | 88 | |
BX640923.1 | - | 3 - 4 | 296 | 88 | |
CR749470.1 | - | 3 - 4 | 317 | 88 | |
KC479043.1 | - | 2 - 3 | 156 | 88 | |
KF435149.1 | - | 2 - 3 | 156 | 88 | |
KF435150.1 | - | 2 - 3 | 156 | 88 |