The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277961.1 | NP_001264890.1 | 10 - 11 | 2042 | 76 |
NM_001277962.1 | NP_001264891.1 | 10 - 11 | 2042 | 76 | |
NM_003156.3 | NP_003147.2 | 10 - 11 | 2042 | 76 | |
GenBank mRNA | AK310510.1 | - | 7 - 8 | 1168 | 76 |
AK314928.1 | - | 10 - 11 | 1563 | 76 | |
BC021300.2 | - | 10 - 11 | 2034 | 76 | |
HQ317451.1 | - | 10 - 11 | 2041 | 76 | |
JX014264.1 | - | 10 - 11 | 1474 | 76 | |
U52426.1 | - | 10 - 11 | 2039 | 76 |