The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003185.3 | NP_003176.2 | 9 - 10 | 2485 | 107 |
GenBank mRNA | AK123937.1 | - | 5 - 6 | 1334 | 107 |
U75308.1 | - | 11 - 12 | 2071 | 107 | |
Y11354.1 | - | 9 - 10 | 2479 | 107 |