The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000405.4 | NP_000396.2 | 1 - 2 | 251 | 87 |
NM_001167607.1 | NP_001161079.1 | 1 - 2 | 251 | 87 | |
GenBank mRNA | AK298867.1 | - | 1 - 2 | 118 | 87 |
BC009273.2 | - | 1 - 2 | 115 | 87 | |
BM834780.1 | - | 1 - 2 | 411 | 87 | |
BX473154.1 | - | 1 - 2 | 229 | 87 | |
L01439.1 | - | 1 - 2 | 176 | 87 | |
M76477.1 | - | 1 - 2 | 176 | 87 | |
X61095.1 | - | 1 - 2 | 107 | 87 | |
X62078.1 | - | 1 - 2 | 144 | 87 |