The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000557.4 | NP_000548.2 | 1 - 2 | 896 | 74 |
NM_001319138.1 | NP_001306067.1 | 3 - 4 | 1138 | 74 | |
XM_011529075.2 | XP_011527377.1 | 3 - 4 | 1028 | 74 | |
GenBank mRNA | AB593125.1 | - | 1 - 2 | 884 | 74 |
BC032495.1 | - | 3 - 4 | 1138 | 74 | |
U13660.1 | - | 1 - 2 | 896 | 74 |