Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTNA
Entrez Gene ID
1837
Gene Name
dystrobrevin alpha
Gene Aliases
D18S892E, DRP3, DTN, DTN-A, LVNC1
Chromosome Location
Chr.18: 34493290 - 34891844 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198938.1 NP_001185867.1 13 - 14 1617 84
NM_001198939.1 NP_001185868.1 13 - 14 1617 84
NM_001198940.1 NP_001185869.1 13 - 14 1617 84
NM_001198941.1 NP_001185870.1 12 - 13 1485 84
NM_001198942.1 NP_001185871.1 8 - 9 1020 84
NM_001198943.1 NP_001185872.1 5 - 6 763 84
NM_001198944.1 NP_001185873.1 6 - 7 849 84
NM_001198945.1 NP_001185874.1 8 - 9 1051 84
NM_001390.4 NP_001381.2 14 - 15 1451 84
NM_001391.5 NP_001382.2 14 - 15 1451 84
NM_032975.3 NP_116757.2 14 - 15 1630 84
NM_032978.6 NP_116760.2 14 - 15 1656 84
NM_032979.4 NP_116761.2 14 - 15 1630 84
NM_032980.3 NP_116762.2 6 - 7 782 84
NM_032981.4 NP_116763.1 5 - 6 704 84
XM_005258221.4 XP_005258278.1 14 - 15 1706 84
XM_011525853.2 XP_011524155.1 13 - 14 1616 84
XM_017025575.1 XP_016881064.1 13 - 14 1575 84
XM_017025576.1 XP_016881065.1 14 - 15 1619 84
XM_017025577.1 XP_016881066.1 12 - 13 1485 84
XM_017025578.1 XP_016881067.1 14 - 15 1700 84
XM_017025579.1 XP_016881068.1 13 - 14 1575 84
XM_017025580.1 XP_016881069.1 14 - 15 1706 84
XM_017025581.1 XP_016881070.1 14 - 15 1700 84
XM_017025582.1 XP_016881071.1 13 - 14 1575 84
XM_017025583.1 XP_016881072.1 14 - 15 1703 84
XM_017025584.1 XP_016881073.1 14 - 15 1619 84
XM_017025585.1 XP_016881074.1 14 - 15 1616 84
XM_017025586.1 XP_016881075.1 14 - 15 1616 84
XM_017025587.1 XP_016881076.1 13 - 14 1610 84
XM_017025588.1 XP_016881077.1 12 - 13 1485 84
XM_017025589.1 XP_016881078.1 13 - 14 1616 84
XM_017025590.1 XP_016881079.1 13 - 14 1610 84
XM_017025591.1 XP_016881080.1 12 - 13 1485 84
XM_017025592.1 XP_016881081.1 13 - 14 1613 84
XM_017025593.1 XP_016881082.1 13 - 14 1607 84
XM_017025594.1 XP_016881083.1 12 - 13 1482 84
XM_017025595.1 XP_016881084.1 14 - 15 1706 84
XM_017025596.1 XP_016881085.1 13 - 14 1613 84
XM_017025597.1 XP_016881086.1 13 - 14 1607 84
XM_017025598.1 XP_016881087.1 12 - 13 1482 84
XM_017025599.1 XP_016881088.1 5 - 6 678 84
GenBank mRNA AB209102.1 - 13 - 14 1570 84
AK291156.1 - 12 - 13 1420 84
AK294746.1 - 8 - 9 1037 84
AK295732.1 - 8 - 9 923 84
AK295789.1 - 6 - 7 740 84
AK296450.1 - 5 - 6 764 84
AK308410.1 - 13 - 14 1597 84
AK315215.1 - 5 - 6 595 84
AL833285.1 - 13 - 14 1577 84
BI668892.1 - 5 - 6 681 84
FJ535564.1 - 8 - 9 724 84
FJ535565.1 - 6 - 7 553 84
U26743.1 - 5 - 6 651 84
U26744.1 - 13 - 14 1337 84
U46744.1 - 13 - 14 1343 84
U46745.1 - 14 - 15 1712 84
U46746.1 - 6 - 7 732 84

View Full Product Details