The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001424.5 | NP_001415.1 | 3 - 4 | 396 | 88 |
XM_006720864.3 | XP_006720927.1 | 3 - 4 | 390 | 88 | |
GenBank mRNA | AK313134.1 | - | 3 - 4 | 312 | 88 |
BC009687.1 | - | 3 - 4 | 287 | 88 | |
BF981157.1 | - | 3 - 4 | 378 | 88 | |
U52100.1 | - | 2 - 3 | 231 | 88 | |
X94770.1 | - | 2 - 3 | 178 | 88 |