Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LPAR1
- Entrez Gene ID
-
1902
- Gene Name
- lysophosphatidic acid receptor 1
- Gene Aliases
-
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
- Chromosome Location
-
Chr.9: 110873252 - 111039259 on Build GRCh38
View Full Product Details