Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
Chromosome Location
Chr.8: 86573608 - 86743675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019098.4 NP_061971.3 4 - 5 540 110
XM_011517138.2 XP_011515440.1 2 - 3 195 110
GenBank mRNA AF228520.1 - 1 - 2 153 110
AF272900.1 - 4 - 5 538 110
BC150601.1 - 4 - 5 518 110

View Full Product Details