The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002736.2 | NP_002727.2 | 4 - 5 | 659 | 93 |
GenBank mRNA | AK128639.1 | - | 5 - 6 | 469 | 93 |
AL559918.3 | - | 4 - 5 | 737 | 93 | |
BC075800.1 | - | 4 - 5 | 698 | 93 | |
CF593944.1 | - | - | 464 | 93 | |
M31158.1 | - | 4 - 5 | 650 | 93 |