The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278273.1 | NP_001265202.1 | 3 - 4 | 220 | 84 |
NM_182691.2 | NP_872633.1 | 1 - 2 | 273 | 84 | |
XM_005250551.4 | XP_005250608.1 | 3 - 4 | 248 | 84 | |
XM_006716098.2 | XP_006716161.1 | 3 - 4 | 215 | 84 | |
XM_011516538.2 | XP_011514840.1 | 2 - 3 | 84 | 84 | |
XM_017012560.1 | XP_016868049.1 | 3 - 4 | 270 | 84 | |
XM_017012561.1 | XP_016868050.1 | 2 - 3 | 120 | 84 | |
XM_017012564.1 | XP_016868053.1 | 1 - 2 | 259 | 84 | |
XM_017012565.1 | XP_016868054.1 | 3 - 4 | 219 | 84 | |
XM_017012566.1 | XP_016868055.1 | 3 - 4 | 256 | 84 | |
GenBank mRNA | AY354201.1 | - | 2 - 3 | 261 | 84 |
BC035214.1 | - | 1 - 2 | 192 | 84 | |
BC068547.1 | - | 3 - 4 | 220 | 84 | |
BM805927.1 | - | - | 247 | 84 | |
DA592321.1 | - | 1 - 2 | 273 | 84 | |
U88666.1 | - | 1 - 2 | 226 | 84 |