The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004073.3 | NP_004064.2 | 5 - 6 | 896 | 77 |
GenBank mRNA | AB209875.1 | - | 6 - 7 | 583 | 77 |
AJ293866.1 | - | 5 - 6 | 896 | 77 | |
BC013899.2 | - | 5 - 6 | 757 | 77 | |
U56998.1 | - | 5 - 6 | 576 | 77 |