The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014794.2 | NP_001014794.1 | 2 - 3 | 248 | 70 |
NM_001014795.2 | NP_001014795.1 | 1 - 2 | 549 | 70 | |
NM_001278441.1 | NP_001265370.1 | 2 - 3 | 294 | 70 | |
NM_004517.3 | NP_004508.1 | 2 - 3 | 294 | 70 | |
XM_005252904.4 | XP_005252961.1 | 2 - 3 | 255 | 70 | |
XM_011520065.1 | XP_011518367.1 | 2 - 3 | 254 | 70 | |
GenBank mRNA | AK296628.1 | - | 2 - 3 | 215 | 70 |
AK314843.1 | - | 2 - 3 | 229 | 70 | |
BC001554.1 | - | 2 - 3 | 155 | 70 | |
CR407673.1 | - | 1 - 2 | 87 | 70 | |
CR749220.1 | - | 1 - 2 | 543 | 70 | |
EU794625.1 | - | 1 - 2 | 382 | 70 | |
U40282.1 | - | 2 - 3 | 243 | 70 |