The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000409.4 | NP_000400.2 | 3 - 4 | 874 | 67 |
NM_001319061.1 | NP_001305990.1 | 3 - 4 | 1245 | 67 | |
NM_001319062.1 | NP_001305991.1 | 2 - 3 | 698 | 67 | |
XM_011514537.2 | XP_011512839.1 | 3 - 4 | 1135 | 67 | |
XM_011514539.1 | XP_011512841.1 | 3 - 4 | 737 | 67 | |
GenBank mRNA | AK125780.1 | - | 3 - 4 | 1176 | 67 |
AK313865.1 | - | 2 - 3 | 669 | 67 | |
BC031663.1 | - | 3 - 4 | 815 | 67 | |
L36859.1 | - | 1 - 2 | 223 | 67 |