The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000527.4 | NP_000518.1 | 15 - 16 | 2502 | 61 |
NM_001195798.1 | NP_001182727.1 | 15 - 16 | 2502 | 61 | |
NM_001195799.1 | NP_001182728.1 | 14 - 15 | 2379 | 61 | |
NM_001195800.1 | NP_001182729.1 | 13 - 14 | 1998 | 61 | |
NM_001195803.1 | NP_001182732.1 | 13 - 14 | 1968 | 61 | |
GenBank mRNA | AB209409.1 | - | 15 - 16 | 2386 | 61 |
AK295612.1 | - | 14 - 15 | 2038 | 61 | |
AK296312.1 | - | 13 - 14 | 1867 | 61 | |
AK299038.1 | - | 13 - 14 | 1897 | 61 | |
AK300313.1 | - | 14 - 15 | 2278 | 61 | |
AY114155.1 | - | 15 - 16 | 2329 | 61 | |
BC014514.1 | - | 15 - 16 | 2385 | 61 | |
BT007361.1 | - | 15 - 16 | 2315 | 61 |