Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16208803-16209311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1M1
Entrez Gene ID
8907
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
UniGene
Hs.71040
Gene Chromosome Location
Chr.19: 16197854-16235345 on Build GRCh38
Transcripts
NM_001130524.1 NM_032493.3 XM_017027420.1 XM_017027421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5497500 c.482C>T p.S161F Chr.19: 16209113-16209113 on Build GRCh38
6423559 c.487G>T p.G163C Chr.19: 16209118-16209118 on Build GRCh38
5651988 c.400T>G p.Y134D Chr.19: 16209031-16209031 on Build GRCh38
4075150 c.465C>T p.N155N Chr.19: 16209096-16209096 on Build GRCh38
4075151 c.538A>G p.N180D Chr.19: 16209169-16209169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866851225 Chr.19: 16208942-16208942 on Build GRCh38
rs376842191 Chr.19: 16209200-16209200 on Build GRCh38
rs775183463 Chr.19: 16209000-16209000 on Build GRCh38
rs760488719 Chr.19: 16209068-16209068 on Build GRCh38
rs766129580 Chr.19: 16209187-16209187 on Build GRCh38
rs45529938 Chr.19: 16208980-16208980 on Build GRCh38
rs372999997 Chr.19: 16209126-16209126 on Build GRCh38
rs890284604 Chr.19: 16209039-16209039 on Build GRCh38
rs374835622 Chr.19: 16209088-16209088 on Build GRCh38
rs774747966 Chr.19: 16209187-16209188 on Build GRCh38
rs2290803 Chr.19: 16209198-16209198 on Build GRCh38
rs372953378 Chr.19: 16208982-16208982 on Build GRCh38
rs2290802 Chr.19: 16209264-16209264 on Build GRCh38
rs778339498 Chr.19: 16208984-16208984 on Build GRCh38
rs45598933 Chr.19: 16208983-16208983 on Build GRCh38
rs760411906 Chr.19: 16209174-16209174 on Build GRCh38
rs984758771 Chr.19: 16208847-16208847 on Build GRCh38
rs749481709 Chr.19: 16209063-16209065 on Build GRCh38
rs1053093579 Chr.19: 16209272-16209272 on Build GRCh38
rs759304514 Chr.19: 16209063-16209064 on Build GRCh38
rs769368822 Chr.19: 16208991-16208991 on Build GRCh38
rs931999109 Chr.19: 16209282-16209282 on Build GRCh38
rs981827489 Chr.19: 16208867-16208867 on Build GRCh38
rs768068780 Chr.19: 16209141-16209141 on Build GRCh38
rs758529330 Chr.19: 16209204-16209204 on Build GRCh38
rs540318197 Chr.19: 16209186-16209186 on Build GRCh38
rs772037968 Chr.19: 16209159-16209159 on Build GRCh38
rs928813856 Chr.19: 16208929-16208929 on Build GRCh38
rs199860370 Chr.19: 16209087-16209087 on Build GRCh38
rs747379798 Chr.19: 16209099-16209099 on Build GRCh38
rs138053047 Chr.19: 16209030-16209030 on Build GRCh38
rs560332890 Chr.19: 16209105-16209105 on Build GRCh38
rs904522004 Chr.19: 16208947-16208947 on Build GRCh38
rs773057955 Chr.19: 16209063-16209063 on Build GRCh38
rs1000225073 Chr.19: 16208952-16208952 on Build GRCh38
rs752864781 Chr.19: 16209203-16209203 on Build GRCh38
rs370207985 Chr.19: 16209224-16209224 on Build GRCh38
rs938881428 Chr.19: 16208938-16208938 on Build GRCh38
rs201575263 Chr.19: 16209016-16209016 on Build GRCh38
rs369323152 Chr.19: 16209098-16209098 on Build GRCh38
rs370707956 Chr.19: 16209202-16209202 on Build GRCh38
rs779606736 Chr.19: 16209122-16209122 on Build GRCh38
rs921920743 Chr.19: 16209278-16209278 on Build GRCh38
rs376188177 Chr.19: 16208990-16208990 on Build GRCh38
rs373845816 Chr.19: 16209072-16209072 on Build GRCh38
rs908768811 Chr.19: 16208862-16208862 on Build GRCh38
rs932851853 Chr.19: 16209138-16209138 on Build GRCh38
rs766560881 Chr.19: 16209071-16209071 on Build GRCh38
rs11557499 Chr.19: 16209066-16209066 on Build GRCh38
rs199888831 Chr.19: 16209214-16209214 on Build GRCh38
rs140278932 Chr.19: 16209168-16209168 on Build GRCh38
rs773112928 Chr.19: 16209165-16209165 on Build GRCh38
rs755015923 Chr.19: 16209077-16209077 on Build GRCh38
rs73928762 Chr.19: 16208943-16208943 on Build GRCh38
rs759036451 Chr.19: 16209096-16209096 on Build GRCh38
rs371056005 Chr.19: 16209010-16209010 on Build GRCh38
rs142277171 Chr.19: 16209215-16209215 on Build GRCh38
rs1049837088 Chr.19: 16208945-16208945 on Build GRCh38
rs940245777 Chr.19: 16209097-16209097 on Build GRCh38
rs763245565 Chr.19: 16209265-16209265 on Build GRCh38
rs891683761 Chr.19: 16208964-16208964 on Build GRCh38
rs1032108651 Chr.19: 16208957-16208957 on Build GRCh38
rs781563340 Chr.19: 16209222-16209222 on Build GRCh38
rs947580384 Chr.19: 16209230-16209230 on Build GRCh38
rs367873999 Chr.19: 16209015-16209015 on Build GRCh38
rs143519787 Chr.19: 16209037-16209037 on Build GRCh38
rs747653326 Chr.19: 16209153-16209153 on Build GRCh38
rs1053360285 Chr.19: 16208941-16208941 on Build GRCh38
rs773912765 Chr.19: 16209148-16209148 on Build GRCh38

View Full Product Details