The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012139.3 | NP_036271.1 | 10 - 11 | 1203 | 61 |
NR_104040.1 | 11 - 12 | 1288 | 61 | ||
NR_104041.1 | 9 - 10 | 1036 | 61 | ||
GenBank mRNA | AJ243950.1 | - | 10 - 11 | 1069 | 61 |
AJ243951.1 | - | 9 - 10 | 902 | 61 | |
AK292286.1 | - | 10 - 11 | 1203 | 61 | |
BC000707.2 | - | 11 - 12 | 1162 | 61 | |
BC065375.1 | - | 10 - 11 | 1183 | 61 |