Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45212506-45213049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L2
Entrez Gene ID
90332
Gene Name
exocyst complex component 3 like 2
Gene Aliases
XTP7
UniGene
Hs.337557
Gene Chromosome Location
Chr.19: 45212621-45234211 on Build GRCh38
Transcripts
NM_138568.3

SNP Detail

SNP ID Chromosome Location
rs370687272 Chr.19: 45212772-45212772 on Build GRCh38
rs902134118 Chr.19: 45213014-45213014 on Build GRCh38
rs1048871284 Chr.19: 45212866-45212866 on Build GRCh38
rs567303856 Chr.19: 45212750-45212750 on Build GRCh38
rs952204099 Chr.19: 45212743-45212743 on Build GRCh38
rs985913542 Chr.19: 45213002-45213002 on Build GRCh38
rs986790638 Chr.19: 45212560-45212560 on Build GRCh38
rs964067918 Chr.19: 45212589-45212589 on Build GRCh38
rs555630174 Chr.19: 45212889-45212889 on Build GRCh38
rs919155376 Chr.19: 45212839-45212839 on Build GRCh38
rs562987294 Chr.19: 45212532-45212532 on Build GRCh38
rs530736333 Chr.19: 45212599-45212601 on Build GRCh38
rs946534566 Chr.19: 45212694-45212694 on Build GRCh38
rs897831290 Chr.19: 45212951-45212951 on Build GRCh38
rs143393432 Chr.19: 45212738-45212738 on Build GRCh38
rs765640635 Chr.19: 45212865-45212865 on Build GRCh38
rs887895883 Chr.19: 45212869-45212869 on Build GRCh38
rs181244785 Chr.19: 45212582-45212582 on Build GRCh38
rs1052262138 Chr.19: 45212655-45212655 on Build GRCh38
rs952313090 Chr.19: 45212545-45212545 on Build GRCh38
rs1003268719 Chr.19: 45212916-45212916 on Build GRCh38
rs1007703298 Chr.19: 45212745-45212745 on Build GRCh38
rs767857620 Chr.19: 45212878-45212878 on Build GRCh38
rs1003975254 Chr.19: 45212701-45212701 on Build GRCh38
rs922386323 Chr.19: 45212598-45212599 on Build GRCh38
rs993579455 Chr.19: 45212729-45212729 on Build GRCh38
rs967822696 Chr.19: 45212749-45212749 on Build GRCh38
rs751688545 Chr.19: 45212600-45212602 on Build GRCh38
rs954321817 Chr.19: 45212976-45212976 on Build GRCh38
rs11667509 Chr.19: 45212934-45212934 on Build GRCh38
rs944345387 Chr.19: 45212785-45212785 on Build GRCh38
rs541830765 Chr.19: 45212698-45212698 on Build GRCh38
rs753637489 Chr.19: 45212599-45212602 on Build GRCh38
rs561698602 Chr.19: 45212733-45212733 on Build GRCh38
rs1031957059 Chr.19: 45212926-45212926 on Build GRCh38
rs976859479 Chr.19: 45212782-45212782 on Build GRCh38
rs574743092 Chr.19: 45212695-45212695 on Build GRCh38
rs11667430 Chr.19: 45212939-45212939 on Build GRCh38
rs114087746 Chr.19: 45212898-45212898 on Build GRCh38
rs1049193330 Chr.19: 45212960-45212960 on Build GRCh38
rs929167542 Chr.19: 45212860-45212860 on Build GRCh38
rs1021806174 Chr.19: 45212951-45212951 on Build GRCh38
rs535422352 Chr.19: 45212811-45212811 on Build GRCh38
rs752310105 Chr.19: 45212890-45212890 on Build GRCh38
rs1022962666 Chr.19: 45212590-45212590 on Build GRCh38
rs3803906 Chr.19: 45212718-45212718 on Build GRCh38
rs1003133794 Chr.19: 45212618-45212618 on Build GRCh38
rs572206227 Chr.19: 45212899-45212899 on Build GRCh38
rs926560152 Chr.19: 45212910-45212910 on Build GRCh38
rs185860487 Chr.19: 45213025-45213025 on Build GRCh38
rs929258112 Chr.19: 45212952-45212952 on Build GRCh38
rs1027717911 Chr.19: 45212528-45212528 on Build GRCh38
rs910227976 Chr.19: 45212769-45212769 on Build GRCh38
rs531937893 Chr.19: 45212562-45212562 on Build GRCh38
rs935076204 Chr.19: 45212598-45212598 on Build GRCh38
rs1021629332 Chr.19: 45212701-45212718 on Build GRCh38
rs971765457 Chr.19: 45212599-45212599 on Build GRCh38
rs910273179 Chr.19: 45213009-45213009 on Build GRCh38
rs577841363 Chr.19: 45213010-45213010 on Build GRCh38
rs966307569 Chr.19: 45212778-45212778 on Build GRCh38
rs369027602 Chr.19: 45212687-45212687 on Build GRCh38
rs976427215 Chr.19: 45212590-45212590 on Build GRCh38

View Full Product Details