Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45224686-45225181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L2
Entrez Gene ID
90332
Gene Name
exocyst complex component 3 like 2
Gene Aliases
XTP7
UniGene
Hs.337557
Gene Chromosome Location
Chr.19: 45212621-45234211 on Build GRCh38
Transcripts
NM_138568.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3103879 c.446G>A p.S149N Chr.19: 45224872-45224872 on Build GRCh38
4454248 c.443A>T p.E148V Chr.19: 45224875-45224875 on Build GRCh38
6368089 c.406G>A p.A136T Chr.19: 45224912-45224912 on Build GRCh38
5973016 c.428G>A p.R143Q Chr.19: 45224890-45224890 on Build GRCh38
5783674 c.521T>G p.L174R Chr.19: 45224797-45224797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758025724 Chr.19: 45224959-45224959 on Build GRCh38
rs376475216 Chr.19: 45224871-45224871 on Build GRCh38
rs752640107 Chr.19: 45224864-45224864 on Build GRCh38
rs373316474 Chr.19: 45224753-45224753 on Build GRCh38
rs917714886 Chr.19: 45225087-45225087 on Build GRCh38
rs1006682540 Chr.19: 45224781-45224781 on Build GRCh38
rs1037823688 Chr.19: 45225151-45225151 on Build GRCh38
rs200984629 Chr.19: 45224831-45224831 on Build GRCh38
rs781258739 Chr.19: 45224788-45224788 on Build GRCh38
rs763933586 Chr.19: 45224907-45224907 on Build GRCh38
rs780283148 Chr.19: 45224861-45224861 on Build GRCh38
rs10411743 Chr.19: 45224980-45224980 on Build GRCh38
rs10410003 Chr.19: 45224865-45224865 on Build GRCh38
rs970447116 Chr.19: 45224745-45224745 on Build GRCh38
rs913532851 Chr.19: 45224843-45224843 on Build GRCh38
rs145422378 Chr.19: 45224812-45224812 on Build GRCh38
rs1047662972 Chr.19: 45225028-45225028 on Build GRCh38
rs779247295 Chr.19: 45224775-45224775 on Build GRCh38
rs966757096 Chr.19: 45225049-45225049 on Build GRCh38
rs10412154 Chr.19: 45225148-45225148 on Build GRCh38
rs750443314 Chr.19: 45224777-45224777 on Build GRCh38
rs148752580 Chr.19: 45224835-45224835 on Build GRCh38
rs565124828 Chr.19: 45224747-45224747 on Build GRCh38
rs35271244 Chr.19: 45224988-45224988 on Build GRCh38
rs766514950 Chr.19: 45224904-45224904 on Build GRCh38
rs750711312 Chr.19: 45224849-45224849 on Build GRCh38
rs10411314 Chr.19: 45224801-45224801 on Build GRCh38
rs775909211 Chr.19: 45224814-45224814 on Build GRCh38
rs933190598 Chr.19: 45224870-45224870 on Build GRCh38
rs202169742 Chr.19: 45224900-45224900 on Build GRCh38
rs764394851 Chr.19: 45224825-45224825 on Build GRCh38
rs541384343 Chr.19: 45225154-45225154 on Build GRCh38
rs574485558 Chr.19: 45224938-45224938 on Build GRCh38
rs746573996 Chr.19: 45224888-45224888 on Build GRCh38
rs968845180 Chr.19: 45224953-45224953 on Build GRCh38
rs775005253 Chr.19: 45224879-45224879 on Build GRCh38
rs397961162 Chr.19: 45225101-45225102 on Build GRCh38
rs959272754 Chr.19: 45225077-45225077 on Build GRCh38
rs1026113636 Chr.19: 45224714-45224714 on Build GRCh38
rs1047613465 Chr.19: 45225112-45225112 on Build GRCh38
rs773236763 Chr.19: 45224901-45224901 on Build GRCh38
rs777456650 Chr.19: 45224807-45224807 on Build GRCh38
rs546141963 Chr.19: 45224808-45224808 on Build GRCh38
rs370054578 Chr.19: 45224878-45224878 on Build GRCh38
rs950788026 Chr.19: 45224722-45224722 on Build GRCh38
rs886527077 Chr.19: 45224749-45224749 on Build GRCh38
rs147446751 Chr.19: 45224796-45224796 on Build GRCh38
rs557776339 Chr.19: 45224872-45224872 on Build GRCh38
rs151207947 Chr.19: 45224890-45224890 on Build GRCh38
rs763296553 Chr.19: 45224903-45224903 on Build GRCh38
rs1030628863 Chr.19: 45225003-45225003 on Build GRCh38
rs1001941007 Chr.19: 45224911-45224911 on Build GRCh38
rs901765202 Chr.19: 45224867-45224867 on Build GRCh38
rs749004945 Chr.19: 45224866-45224866 on Build GRCh38
rs758622299 Chr.19: 45224784-45224784 on Build GRCh38
rs141848499 Chr.19: 45225024-45225024 on Build GRCh38
rs199986766 Chr.19: 45224935-45224935 on Build GRCh38
rs753870772 Chr.19: 45224910-45224910 on Build GRCh38
rs757445762 Chr.19: 45224767-45224767 on Build GRCh38
rs960292322 Chr.19: 45224934-45224934 on Build GRCh38
rs537583453 Chr.19: 45224813-45224813 on Build GRCh38
rs949316321 Chr.19: 45225108-45225108 on Build GRCh38
rs1013530251 Chr.19: 45224721-45224721 on Build GRCh38
rs1006283103 Chr.19: 45224727-45224727 on Build GRCh38
rs192440405 Chr.19: 45225085-45225085 on Build GRCh38
rs533302660 Chr.19: 45224757-45224757 on Build GRCh38
rs545167719 Chr.19: 45225058-45225058 on Build GRCh38
rs368599925 Chr.19: 45225040-45225040 on Build GRCh38
rs57260076 Chr.19: 45225097-45225098 on Build GRCh38
rs138255702 Chr.19: 45225014-45225014 on Build GRCh38
rs147432288 Chr.19: 45224965-45224965 on Build GRCh38
rs145255138 Chr.19: 45224824-45224824 on Build GRCh38
rs571153044 Chr.19: 45224898-45224898 on Build GRCh38
rs979238988 Chr.19: 45224957-45224957 on Build GRCh38
rs10410561 Chr.19: 45224973-45224973 on Build GRCh38
rs755147222 Chr.19: 45224905-45224905 on Build GRCh38
rs765643980 Chr.19: 45224899-45224899 on Build GRCh38
rs765377183 Chr.19: 45224848-45224848 on Build GRCh38
rs886303943 Chr.19: 45225054-45225054 on Build GRCh38
rs1035619835 Chr.19: 45224931-45224931 on Build GRCh38
rs1016287677 Chr.19: 45224728-45224728 on Build GRCh38
rs925014369 Chr.19: 45225066-45225066 on Build GRCh38
rs745475883 Chr.19: 45224947-45224947 on Build GRCh38
rs777265731 Chr.19: 45224892-45224892 on Build GRCh38
rs569002007 Chr.19: 45224860-45224860 on Build GRCh38
rs942013266 Chr.19: 45225150-45225150 on Build GRCh38
rs753261271 Chr.19: 45224738-45224738 on Build GRCh38
rs770466337 Chr.19: 45224810-45224810 on Build GRCh38

View Full Product Details