Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45549849-45550391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPA3
Entrez Gene ID
80207
Gene Name
optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia)
Gene Aliases
MGA3
UniGene
Hs.466945
Gene Chromosome Location
Chr.19: 45527767-45584864 on Build GRCh38
Transcripts
NM_001017989.2 NM_025136.3 XM_006723403.3

SNP Detail

SNP ID Chromosome Location
rs1004348319 Chr.19: 45550278-45550278 on Build GRCh38
rs545764728 Chr.19: 45550154-45550154 on Build GRCh38
rs1021505529 Chr.19: 45550332-45550332 on Build GRCh38
rs985196664 Chr.19: 45549928-45549928 on Build GRCh38
rs1048679986 Chr.19: 45550201-45550201 on Build GRCh38
rs561119752 Chr.19: 45550120-45550120 on Build GRCh38
rs894456563 Chr.19: 45550212-45550212 on Build GRCh38
rs1048647700 Chr.19: 45550017-45550017 on Build GRCh38
rs576070088 Chr.19: 45549933-45549933 on Build GRCh38
rs944230445 Chr.19: 45549934-45549934 on Build GRCh38
rs918763163 Chr.19: 45550161-45550161 on Build GRCh38
rs1002749763 Chr.19: 45550069-45550069 on Build GRCh38
rs764494903 Chr.19: 45550307-45550307 on Build GRCh38
rs553328047 Chr.19: 45550075-45550075 on Build GRCh38
rs955986711 Chr.19: 45550102-45550102 on Build GRCh38
rs541235547 Chr.19: 45550107-45550107 on Build GRCh38
rs770317628 Chr.19: 45550236-45550236 on Build GRCh38
rs142831259 Chr.19: 45550194-45550194 on Build GRCh38
rs530530814 Chr.19: 45550337-45550337 on Build GRCh38
rs1011327136 Chr.19: 45550314-45550314 on Build GRCh38
rs926034254 Chr.19: 45550179-45550179 on Build GRCh38
rs946136123 Chr.19: 45549937-45549937 on Build GRCh38
rs1046483724 Chr.19: 45550051-45550051 on Build GRCh38
rs1037601092 Chr.19: 45549955-45549955 on Build GRCh38
rs768641413 Chr.19: 45550071-45550071 on Build GRCh38
rs3760844 Chr.19: 45550127-45550127 on Build GRCh38
rs907572933 Chr.19: 45549885-45549885 on Build GRCh38
rs181719786 Chr.19: 45550164-45550164 on Build GRCh38
rs916799566 Chr.19: 45550221-45550221 on Build GRCh38
rs1029106202 Chr.19: 45550330-45550330 on Build GRCh38
rs973056688 Chr.19: 45549898-45549898 on Build GRCh38
rs1056216871 Chr.19: 45550210-45550210 on Build GRCh38
rs1014481210 Chr.19: 45550043-45550043 on Build GRCh38
rs558779723 Chr.19: 45550052-45550052 on Build GRCh38
rs936070897 Chr.19: 45550188-45550192 on Build GRCh38
rs931537597 Chr.19: 45549984-45549984 on Build GRCh38
rs965480314 Chr.19: 45550156-45550156 on Build GRCh38
rs921845380 Chr.19: 45549900-45549900 on Build GRCh38
rs953958991 Chr.19: 45550366-45550366 on Build GRCh38
rs73568980 Chr.19: 45550292-45550292 on Build GRCh38
rs189068529 Chr.19: 45550138-45550138 on Build GRCh38
rs373580645 Chr.19: 45550157-45550161 on Build GRCh38
rs79317386 Chr.19: 45550302-45550302 on Build GRCh38
rs921593113 Chr.19: 45550153-45550153 on Build GRCh38
rs112899014 Chr.19: 45549983-45549983 on Build GRCh38
rs942748678 Chr.19: 45549935-45549935 on Build GRCh38
rs772710835 Chr.19: 45550157-45550157 on Build GRCh38
rs541630766 Chr.19: 45549923-45549923 on Build GRCh38
rs138449967 Chr.19: 45550072-45550072 on Build GRCh38
rs963014773 Chr.19: 45549896-45549896 on Build GRCh38
rs894738657 Chr.19: 45550040-45550041 on Build GRCh38
rs559474405 Chr.19: 45550350-45550350 on Build GRCh38
rs749084024 Chr.19: 45550018-45550018 on Build GRCh38
rs147400347 Chr.19: 45550225-45550225 on Build GRCh38
rs973918302 Chr.19: 45549946-45549946 on Build GRCh38
rs1012214154 Chr.19: 45550020-45550020 on Build GRCh38
rs139795580 Chr.19: 45550352-45550352 on Build GRCh38
rs113095552 Chr.19: 45550338-45550338 on Build GRCh38
rs931892968 Chr.19: 45549906-45549906 on Build GRCh38
rs887313631 Chr.19: 45550243-45550243 on Build GRCh38
rs185631253 Chr.19: 45550234-45550234 on Build GRCh38
rs577805330 Chr.19: 45550348-45550348 on Build GRCh38
rs911715259 Chr.19: 45549924-45549924 on Build GRCh38

View Full Product Details