The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005336.2 | NP_001005336.1 | 16 - 17 | 1898 | 66 |
NM_001288737.1 | NP_001275666.1 | 16 - 17 | 1898 | 66 | |
NM_001288738.1 | NP_001275667.1 | 16 - 17 | 1898 | 66 | |
NM_001288739.1 | NP_001275668.1 | 16 - 17 | 1898 | 66 | |
NM_004408.3 | NP_004399.2 | 16 - 17 | 1898 | 66 | |
XM_005251763.2 | XP_005251820.1 | 16 - 17 | 1898 | 66 | |
XM_005251768.2 | XP_005251825.1 | 16 - 17 | 1898 | 66 | |
XM_005251769.2 | XP_005251826.1 | 16 - 17 | 1898 | 66 | |
XM_006716992.2 | XP_006717055.1 | 16 - 17 | 1898 | 66 | |
XM_011518335.2 | XP_011516637.1 | 15 - 16 | 1886 | 66 | |
XM_011518336.2 | XP_011516638.1 | 15 - 16 | 1886 | 66 | |
XM_017014369.1 | XP_016869858.1 | 16 - 17 | 1898 | 66 | |
XM_017014370.1 | XP_016869859.1 | 16 - 17 | 1898 | 66 | |
XM_017014371.1 | XP_016869860.1 | 16 - 17 | 1898 | 66 | |
XM_017014373.1 | XP_016869862.1 | 15 - 16 | 1886 | 66 | |
GenBank mRNA | AB209124.1 | - | 15 - 16 | 1583 | 66 |
AK096629.1 | - | 6 - 7 | 3088 | 66 | |
AK225179.1 | - | 16 - 17 | 1807 | 66 | |
AK295101.1 | - | 16 - 17 | 1691 | 66 | |
AK296326.1 | - | 16 - 17 | 1865 | 66 | |
AK308793.1 | - | 9 - 10 | 699 | 66 | |
AK316235.1 | - | 16 - 17 | 1874 | 66 | |
BC050279.1 | - | 16 - 17 | 1874 | 66 | |
BC063850.1 | - | 16 - 17 | 1831 | 66 | |
L07807.1 | - | 16 - 17 | 1848 | 66 |