Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9584955-9585493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAQ
Entrez Gene ID
10971
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta
Gene Aliases
14-3-3, 1C5, HS1
UniGene
Hs.74405
Gene Chromosome Location
Chr.2: 9583967-9631055 on Build GRCh38
Transcripts
NM_006826.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6824609 c.728C>T p.A243V Chr.2: 9585296-9585296 on Build GRCh38
5796084 c.713A>T p.D238V Chr.2: 9585311-9585311 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs573366066 Chr.2: 9585142-9585142 on Build GRCh38
rs766333650 Chr.2: 9585418-9585418 on Build GRCh38
rs145097891 Chr.2: 9585267-9585267 on Build GRCh38
rs958805932 Chr.2: 9585413-9585413 on Build GRCh38
rs762224503 Chr.2: 9585249-9585249 on Build GRCh38
rs985985030 Chr.2: 9585082-9585082 on Build GRCh38
rs766715734 Chr.2: 9585377-9585377 on Build GRCh38
rs947268963 Chr.2: 9585446-9585446 on Build GRCh38
rs780100582 Chr.2: 9585273-9585273 on Build GRCh38
rs187340849 Chr.2: 9584994-9584994 on Build GRCh38
rs763356905 Chr.2: 9585366-9585366 on Build GRCh38
rs528409502 Chr.2: 9585411-9585411 on Build GRCh38
rs752671424 Chr.2: 9585392-9585392 on Build GRCh38
rs373178591 Chr.2: 9585307-9585307 on Build GRCh38
rs892576436 Chr.2: 9585200-9585200 on Build GRCh38
rs924307077 Chr.2: 9585467-9585467 on Build GRCh38
rs1041615467 Chr.2: 9585460-9585460 on Build GRCh38
rs148399453 Chr.2: 9585289-9585289 on Build GRCh38
rs548374954 Chr.2: 9585448-9585448 on Build GRCh38
rs781125712 Chr.2: 9585283-9585283 on Build GRCh38
rs375133910 Chr.2: 9585375-9585375 on Build GRCh38
rs1040610902 Chr.2: 9585195-9585195 on Build GRCh38
rs753821484 Chr.2: 9585296-9585296 on Build GRCh38
rs779806817 Chr.2: 9585311-9585311 on Build GRCh38
rs369470995 Chr.2: 9585260-9585260 on Build GRCh38
rs928718171 Chr.2: 9585370-9585370 on Build GRCh38
rs775943040 Chr.2: 9585328-9585328 on Build GRCh38
rs375784718 Chr.2: 9585265-9585265 on Build GRCh38
rs200376025 Chr.2: 9585308-9585308 on Build GRCh38
rs752577017 Chr.2: 9585321-9585323 on Build GRCh38
rs777255377 Chr.2: 9585240-9585240 on Build GRCh38
rs750627405 Chr.2: 9585176-9585176 on Build GRCh38
rs766682675 Chr.2: 9585261-9585263 on Build GRCh38
rs746631800 Chr.2: 9585315-9585315 on Build GRCh38
rs757292325 Chr.2: 9585393-9585393 on Build GRCh38
rs764110865 Chr.2: 9585394-9585400 on Build GRCh38
rs762328022 Chr.2: 9585329-9585329 on Build GRCh38
rs112834388 Chr.2: 9585430-9585431 on Build GRCh38
rs767650830 Chr.2: 9585391-9585391 on Build GRCh38
rs16822436 Chr.2: 9585194-9585194 on Build GRCh38
rs41264177 Chr.2: 9585266-9585266 on Build GRCh38
rs191830707 Chr.2: 9585268-9585268 on Build GRCh38
rs910377629 Chr.2: 9585097-9585097 on Build GRCh38
rs751873357 Chr.2: 9585381-9585381 on Build GRCh38
rs534969469 Chr.2: 9585405-9585405 on Build GRCh38
rs76429341 Chr.2: 9585117-9585117 on Build GRCh38
rs376453415 Chr.2: 9585239-9585239 on Build GRCh38
rs374886445 Chr.2: 9585087-9585087 on Build GRCh38
rs944592786 Chr.2: 9585188-9585188 on Build GRCh38
rs199737672 Chr.2: 9585286-9585286 on Build GRCh38
rs1027292482 Chr.2: 9585400-9585402 on Build GRCh38
rs914484072 Chr.2: 9585437-9585437 on Build GRCh38
rs757206430 Chr.2: 9585300-9585300 on Build GRCh38
rs755141288 Chr.2: 9585281-9585281 on Build GRCh38
rs574721044 Chr.2: 9585008-9585008 on Build GRCh38
rs112750534 Chr.2: 9585025-9585025 on Build GRCh38
rs373509981 Chr.2: 9585390-9585390 on Build GRCh38
rs184806272 Chr.2: 9585356-9585356 on Build GRCh38
rs768281092 Chr.2: 9585325-9585325 on Build GRCh38
rs770242992 Chr.2: 9585355-9585355 on Build GRCh38

View Full Product Details