Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9591311-9591786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAQ
Entrez Gene ID
10971
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta
Gene Aliases
14-3-3, 1C5, HS1
UniGene
Hs.74405
Gene Chromosome Location
Chr.2: 9583967-9631055 on Build GRCh38
Transcripts
NM_006826.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6824613 c.329C>G p.T110S Chr.2: 9591481-9591481 on Build GRCh38
286607 c.319G>T p.A107S Chr.2: 9591491-9591491 on Build GRCh38
6824611 c.418C>T p.Q140* Chr.2: 9591392-9591392 on Build GRCh38
5973946 c.396T>G p.V132V Chr.2: 9591414-9591414 on Build GRCh38
6824610 c.410A>C p.D137A Chr.2: 9591400-9591400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756536691 Chr.2: 9591375-9591375 on Build GRCh38
rs185211853 Chr.2: 9591482-9591482 on Build GRCh38
rs189633973 Chr.2: 9591747-9591747 on Build GRCh38
rs73913140 Chr.2: 9591760-9591760 on Build GRCh38
rs764670151 Chr.2: 9591539-9591539 on Build GRCh38
rs756461336 Chr.2: 9591347-9591347 on Build GRCh38
rs143350262 Chr.2: 9591598-9591598 on Build GRCh38
rs976100358 Chr.2: 9591752-9591752 on Build GRCh38
rs773284973 Chr.2: 9591411-9591411 on Build GRCh38
rs374812266 Chr.2: 9591633-9591633 on Build GRCh38
rs563403576 Chr.2: 9591631-9591631 on Build GRCh38
rs946892734 Chr.2: 9591477-9591477 on Build GRCh38
rs999803012 Chr.2: 9591597-9591597 on Build GRCh38
rs543810769 Chr.2: 9591611-9591611 on Build GRCh38
rs374320554 Chr.2: 9591541-9591541 on Build GRCh38
rs779650867 Chr.2: 9591347-9591348 on Build GRCh38
rs775375833 Chr.2: 9591487-9591487 on Build GRCh38
rs138977724 Chr.2: 9591646-9591646 on Build GRCh38
rs892847212 Chr.2: 9591476-9591476 on Build GRCh38
rs1055486082 Chr.2: 9591429-9591429 on Build GRCh38
rs961952018 Chr.2: 9591593-9591593 on Build GRCh38
rs780753322 Chr.2: 9591560-9591560 on Build GRCh38
rs376583183 Chr.2: 9591536-9591536 on Build GRCh38
rs749903210 Chr.2: 9591547-9591547 on Build GRCh38
rs768853936 Chr.2: 9591547-9591582 on Build GRCh38
rs769768341 Chr.2: 9591392-9591392 on Build GRCh38
rs763862962 Chr.2: 9591525-9591525 on Build GRCh38
rs57340089 Chr.2: 9591366-9591366 on Build GRCh38
rs892737315 Chr.2: 9591339-9591339 on Build GRCh38
rs112226455 Chr.2: 9591509-9591509 on Build GRCh38
rs962823215 Chr.2: 9591662-9591662 on Build GRCh38
rs753505265 Chr.2: 9591526-9591526 on Build GRCh38
rs747405548 Chr.2: 9591519-9591522 on Build GRCh38
rs368957039 Chr.2: 9591741-9591741 on Build GRCh38
rs760512737 Chr.2: 9591489-9591489 on Build GRCh38
rs1043933165 Chr.2: 9591481-9591481 on Build GRCh38
rs902676908 Chr.2: 9591495-9591495 on Build GRCh38
rs747415430 Chr.2: 9591343-9591343 on Build GRCh38
rs372150851 Chr.2: 9591761-9591761 on Build GRCh38
rs781619470 Chr.2: 9591370-9591370 on Build GRCh38
rs541118173 Chr.2: 9591385-9591385 on Build GRCh38
rs768671602 Chr.2: 9591717-9591717 on Build GRCh38
rs780590064 Chr.2: 9591341-9591341 on Build GRCh38
rs974562287 Chr.2: 9591673-9591673 on Build GRCh38
rs758097076 Chr.2: 9591348-9591348 on Build GRCh38
rs867290853 Chr.2: 9591554-9591554 on Build GRCh38
rs981748435 Chr.2: 9591374-9591374 on Build GRCh38
rs1025501104 Chr.2: 9591679-9591679 on Build GRCh38
rs1023895805 Chr.2: 9591577-9591577 on Build GRCh38
rs759412655 Chr.2: 9591474-9591474 on Build GRCh38
rs757948276 Chr.2: 9591557-9591557 on Build GRCh38

View Full Product Details