The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006267.4 | NP_006258.3 | 18 - 19 | 2726 | 136 |
XM_005264002.2 | XP_005264059.1 | 18 - 19 | 2761 | 136 | |
XM_005264003.2 | XP_005264060.1 | 18 - 19 | 2689 | 136 | |
XM_005264004.2 | XP_005264061.1 | 18 - 19 | 2689 | 136 | |
XM_005264005.4 | XP_005264062.1 | 18 - 19 | 2749 | 136 | |
XM_005264007.2 | XP_005264064.1 | 18 - 19 | 2689 | 136 | |
XM_011511575.2 | XP_011509877.1 | 18 - 19 | 2664 | 136 | |
XM_011511576.2 | XP_011509878.1 | 18 - 19 | 2689 | 136 | |
XM_011511578.2 | XP_011509880.1 | 18 - 19 | 2731 | 136 | |
XM_017004623.1 | XP_016860112.1 | 18 - 19 | 2689 | 136 | |
GenBank mRNA | AB209483.1 | - | 15 - 16 | 2342 | 136 |
AK026993.1 | - | 2 - 3 | 155 | 136 | |
D42063.1 | - | 18 - 19 | 2727 | 136 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024457.3 | NP_001019628.3 | 18 - 19 | 2571 | 136 |
XM_011532845.2 | XP_011531147.1 | 19 - 20 | 2770 | 136 | |
XM_011532849.2 | XP_011531151.1 | 19 - 20 | 2770 | 136 | |
XM_011532850.1 | XP_011531152.1 | 19 - 20 | 2770 | 136 | |
XM_011532851.2 | XP_011531153.1 | 19 - 20 | 2770 | 136 | |
XM_011532852.2 | XP_011531154.1 | 19 - 20 | 2770 | 136 | |
XM_017004103.1 | XP_016859592.1 | 19 - 20 | 2770 | 136 | |
XM_017004104.1 | XP_016859593.1 | 18 - 19 | 2887 | 136 | |
XM_017004105.1 | XP_016859594.1 | 18 - 19 | 2735 | 136 | |
XM_017004106.1 | XP_016859595.1 | 18 - 19 | 2854 | 136 | |
XM_017004107.1 | XP_016859596.1 | 19 - 20 | 2770 | 136 | |
GenBank mRNA | AK303443.1 | - | 4 - 5 | 487 | 136 |
CR749330.1 | - | 18 - 19 | 2571 | 136 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001078170.2 | NP_001071638.2 | 18 - 19 | 2617 | 136 |
XM_017004845.1 | XP_016860334.1 | 22 - 23 | 3068 | 136 | |
XM_017004846.1 | XP_016860335.1 | 18 - 19 | 2739 | 136 | |
XM_017004847.1 | XP_016860336.1 | 18 - 19 | 2736 | 136 | |
XM_017004848.1 | XP_016860337.1 | 22 - 23 | 3068 | 136 | |
XM_017004849.1 | XP_016860338.1 | 22 - 23 | 3068 | 136 | |
XM_017004850.1 | XP_016860339.1 | 22 - 23 | 3068 | 136 | |
GenBank mRNA | AK300352.1 | - | 7 - 8 | 957 | 136 |
AK302429.1 | - | 18 - 19 | 2617 | 136 | |
AY354203.1 | - | 5 - 6 | 593 | 136 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144013.1 | NP_001137485.1 | 18 - 19 | 2660 | 136 |
XM_017004738.1 | XP_016860227.1 | 19 - 20 | 2709 | 136 | |
XM_017004739.1 | XP_016860228.1 | 19 - 20 | 2709 | 136 | |
GenBank mRNA | AK310529.1 | - | 4 - 5 | 439 | 136 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182588.2 | NP_872394.2 | 18 - 19 | 2685 | 136 |
XM_011511017.2 | XP_011509319.1 | 18 - 19 | 2632 | 136 | |
XM_011511018.2 | XP_011509320.1 | 18 - 19 | 2635 | 136 | |
XM_011511019.2 | XP_011509321.1 | 18 - 19 | 2631 | 136 | |
XM_011511020.2 | XP_011509322.1 | 18 - 19 | 2631 | 136 | |
GenBank mRNA | AK302921.1 | - | 14 - 15 | 1993 | 136 |
BX537861.1 | - | 13 - 14 | 3583 | 136 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005054.2 | NP_005045.2 | 18 - 19 | 2677 | 136 |
XM_006712794.3 | XP_006712857.2 | 18 - 19 | 2708 | 136 | |
XM_017005080.1 | XP_016860569.1 | 18 - 19 | 2697 | 136 | |
XM_017005081.1 | XP_016860570.1 | 18 - 19 | 2708 | 136 | |
XM_017005082.1 | XP_016860571.1 | 16 - 17 | 2379 | 136 | |
GenBank mRNA | EU794682.1 | - | 18 - 19 | 2677 | 136 |
U64675.2 | - | 18 - 19 | 2677 | 136 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123363.3 | NP_001116835.1 | 19 - 20 | 3133 | 136 |
XM_011511766.2 | XP_011510068.1 | 18 - 19 | 2717 | 136 | |
XM_011511767.2 | XP_011510069.1 | 18 - 19 | 2717 | 136 | |
XM_011511769.2 | XP_011510071.1 | 18 - 19 | 2717 | 136 | |
XM_011511770.2 | XP_011510072.1 | 18 - 19 | 2717 | 136 | |
XM_017004839.1 | XP_016860328.1 | 18 - 19 | 2717 | 136 | |
XM_017004840.1 | XP_016860329.1 | 18 - 19 | 2790 | 136 | |
XM_017004841.1 | XP_016860330.1 | 18 - 19 | 2920 | 136 | |
XM_017004842.1 | XP_016860331.1 | 18 - 19 | 2697 | 136 | |
XM_017004843.1 | XP_016860332.1 | 18 - 19 | 2717 | 136 | |
XM_017004844.1 | XP_016860333.1 | 16 - 17 | 2379 | 136 | |
GenBank mRNA | AK056675.1 | - | 14 - 15 | 1991 | 136 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164463.1 | NP_001157935.1 | 18 - 19 | 2792 | 136 |
XM_005263747.4 | XP_005263804.1 | 18 - 19 | 2790 | 136 | |
XM_011511733.2 | XP_011510035.1 | 18 - 19 | 2790 | 136 | |
XM_017004825.1 | XP_016860314.1 | 18 - 19 | 2790 | 136 |