The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168398.1 | NP_001161870.1 | 4 - 5 | 582 | 62 |
NM_006416.4 | NP_006407.1 | 4 - 5 | 582 | 62 | |
GenBank mRNA | AJ810302.1 | - | 4 - 5 | 516 | 62 |
AJ810303.1 | - | 4 - 5 | 518 | 62 | |
AJ851888.1 | - | 4 - 5 | 503 | 62 | |
AJ851890.1 | - | 3 - 4 | 343 | 62 | |
AK293696.1 | - | 3 - 4 | 374 | 62 | |
BC017807.1 | - | 4 - 5 | 530 | 62 | |
D87969.1 | - | 4 - 5 | 530 | 62 |