The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278439.1 | NP_001265368.1 | 4 - 5 | 291 | 80 |
NM_007056.2 | NP_008987.2 | 5 - 6 | 477 | 80 | |
NR_103529.1 | 5 - 6 | 477 | 80 | ||
XM_011526396.2 | XP_011524698.1 | 5 - 6 | 472 | 80 | |
XM_011526397.2 | XP_011524699.1 | 4 - 5 | 322 | 80 | |
GenBank mRNA | AF042800.1 | - | 5 - 6 | 477 | 80 |
AK094681.1 | - | 4 - 5 | 1071 | 80 | |
AK293333.1 | - | 4 - 5 | 287 | 80 | |
AY358944.1 | - | 5 - 6 | 458 | 80 | |
BC013178.1 | - | 5 - 6 | 422 | 80 | |
BC080554.1 | - | 4 - 5 | 417 | 80 | |
BC131496.1 | - | 4 - 5 | 335 | 80 |