The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012244.3 | NP_036376.2 | 1 - 2 | 897 | 87 |
GenBank mRNA | AB037669.1 | - | 1 - 2 | 860 | 87 |
AF135828.1 | - | 1 - 2 | 372 | 87 | |
AF171669.1 | - | 1 - 2 | 374 | 87 | |
AK223610.1 | - | 1 - 2 | 896 | 87 | |
AK313465.1 | - | 1 - 2 | 151 | 87 | |
AL365343.2 | - | 1 - 2 | 401 | 87 | |
AL365414.1 | - | 1 - 2 | 401 | 87 | |
BC052250.1 | - | 1 - 2 | 180 | 87 | |
Y18483.1 | - | 1 - 2 | 881 | 87 |