The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013337.2 | NP_037469.2 | 1 - 2 | 259 | 81 |
GenBank mRNA | AF155330.1 | - | 1 - 2 | 238 | 81 |
AK000830.1 | - | 1 - 2 | 240 | 81 | |
AK225453.1 | - | 1 - 2 | 240 | 81 | |
BC002324.2 | - | 1 - 2 | 259 | 81 |