The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013974.2 | NP_039268.1 | 1 - 2 | 212 | 85 |
GenBank mRNA | AF070667.1 | - | 1 - 2 | 209 | 85 |
AF087894.1 | - | 1 - 2 | 209 | 85 | |
AK098671.1 | - | 1 - 2 | 199 | 85 | |
BC001435.2 | - | 1 - 2 | 124 | 85 | |
FJ224290.1 | - | 1 - 2 | 209 | 85 |