Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF544
Entrez Gene ID
27300
Gene Name
zinc finger protein 544
Gene Aliases
-
Chromosome Location
Chr.19: 58228681 - 58277497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320767.1 NP_001307696.1 4 - 5 565 86
NM_001320769.1 NP_001307698.1 4 - 5 607 86
NM_001320770.1 NP_001307699.1 4 - 5 565 86
NM_001320771.1 NP_001307700.1 4 - 5 530 86
NM_001320773.1 NP_001307702.1 4 - 5 607 86
NM_001320774.1 NP_001307703.1 4 - 5 607 86
NM_001320776.1 NP_001307705.1 5 - 6 876 86
NM_001320777.1 NP_001307706.1 5 - 6 876 86
NM_001320780.1 NP_001307709.1 4 - 5 530 86
NM_001320781.1 NP_001307710.1 5 - 6 953 86
NM_001320782.1 NP_001307711.1 4 - 5 530 86
NM_001320783.1 NP_001307712.1 4 - 5 607 86
NM_001320785.1 NP_001307714.1 4 - 5 530 86
NM_001320786.1 NP_001307715.1 4 - 5 565 86
NM_001320787.1 NP_001307716.1 4 - 5 607 86
NM_001320788.1 NP_001307717.1 4 - 5 530 86
NM_001320789.1 NP_001307718.1 4 - 5 607 86
NM_001320791.1 NP_001307720.1 4 - 5 565 86
NM_001320792.1 NP_001307721.1 5 - 6 616 86
NM_014480.3 NP_055295.2 4 - 5 530 86
XM_011526738.2 XP_011525040.1 5 - 6 643 86
XM_017026599.1 XP_016882088.1 5 - 6 643 86
XM_017026600.1 XP_016882089.1 5 - 6 643 86
XM_017026601.1 XP_016882090.1 5 - 6 643 86
XM_017026602.1 XP_016882091.1 5 - 6 643 86
XM_017026603.1 XP_016882092.1 5 - 6 643 86
XM_017026604.1 XP_016882093.1 5 - 6 643 86
XM_017026605.1 XP_016882094.1 5 - 6 643 86
GenBank mRNA AF020591.1 - 3 - 4 278 86
AK226080.1 - 4 - 5 322 86
AK291656.1 - 4 - 5 282 86
AK296846.1 - 3 - 4 275 86
AK301650.1 - 4 - 5 282 86
AK316446.1 - 4 - 5 317 86
AL833618.1 - 4 - 5 294 86
BC042177.1 - 3 - 4 274 86
BC067271.1 - 3 - 4 258 86
CB997427.1 - 4 - 5 542 86
DA342047.1 - 4 - 5 317 86
DB200767.1 - 4 - 5 282 86

View Full Product Details